Vai a
BR782811
BRAND® insert strips for 24 x 6-well BRANDplates®
Inlet Opening System, PET membrane, pore size 0.4 μm
Sinonimo/i:
BRANDplates®, Cell culture, Insert Strips, Insert System, Inserts, Tissue culture
Autenticati per visualizzare i prezzi organizzativi e contrattuali.
Scegli un formato
Cambia visualizzazione
Voi/SKUDisponibilitàPrezzo
12 ea
Per conoscere la disponibilità, visualizza il carrello
CHF 350.00
Informazioni su questo articolo
NACRES:
NB.22
UNSPSC Code:
41122107
Material:
PET membrane
Pore size:
0.4 μm
Sterility:
sterile
CHF 350.00
Per conoscere la disponibilità, visualizza il carrello
​
Servizio Tecnico
Hai bisogno di aiuto? Il nostro team di scienziati qualificati è a tua disposizione.
Permettici di aiutarti
material
PET membrane
description
cellGrade™ plus
sterility
sterile
packaging
pack of 12 ea (strips of 4 inserts, individually wrapped)
manufacturer/tradename
BRAND 782811
size
6 well
pore size
0.4 μm
General description
Innovative cell culture plates and inserts for manual and automatic cell and tissue culture. The BRANDplates® Insert System, developed in conjunction with the Fraunhofer company, offers an innovative expansion to BRAND′s existing cell culture product range. It consists of two different cell culture plates and two designs of cell culture inserts that can be combined in different configurations.Insert specification:4x insert strips, can be divided into up to 4 individual inserts - hanging position in wellHanging inserts – distance between the well base and insert membrane is 4 mmIndividually packaged strips inserted in thermoformed beakers with 6 strips each in the 6-well cell culture platecellGrade™ plus (hydrophilic cell culture-) surface.Smooth wall inserts: For standard applications like secretion studies, co-cultivations, chemotaxis tests, etc.PS cellGrade™ plus surface, sterile, strip of 4 inserts (divisible).BIO-CERT® CELL CULTURE QUALITY: Free of human DNA, RNase, DNase, Pyrogens and non cytotoxic
Legal Information
BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
BRANDplates is a registered trademark of BRAND GMBH + CO KG
cellGrade is a trademark of BRAND GMBH + CO KG
Confronta articoli simili
Visualizza il confronto per intero
Mostra le differenze
1 of 1
Questo articolo
BR782821BR782810BR782861
description
Inlet Opening System, PET membrane, pore size 0.4 μm
description
Inlet Opening System, PC membrane, pore size 1 μm
description
PET membrane, pore size 0.4 μm
description
Inlet Opening System, PC membrane, pore size 8 μm
manufacturer/tradename
BRAND 782811
manufacturer/tradename
BRAND 782821
manufacturer/tradename
BRAND 782810
manufacturer/tradename
BRAND 782861
sterility
sterile
sterility
sterile
sterility
sterile
sterility
sterile
pore size
0.4 μm
pore size
1 μm
pore size
0.4 μm
pore size
8 μm
material
PET membrane
material
PC membrane
material
PET membrane
material
PC membrane
size
6 well
size
6 well
size
6 well
size
6 well
packaging
pack of 12 ea (strips of 4 inserts, individually wrapped)
packaging
pack of 12 ea (strips of 4 inserts, individually wrapped)
packaging
pack of 12 ea (strips of 4 inserts, individually wrapped)
packaging
pack of 12 ea (strips of 4 inserts, individually wrapped)
Still not finding the right product?
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)