Scegli un formato
| Voi/SKU | Disponibilità | Prezzo |
|---|
Informazioni su questo articolo
material
silicone (research-grade)
sterility
non-sterile
manufacturer/tradename
Axygen Inc AM-2ML-RD-IMP, Corning
packaging
pkg of 5x10mats/cs
size
96 wells
1 of 1
Questo articolo | |||
|---|---|---|---|
| description for 2 mL round deep well plates, size 96 wells | description for 2 mL square deep well plates, size 96 wells | description for 2 mL square deep well plates, size 96 wells, sterile | description for 5 mL rectangular deep well plates, size 48 wells |
| manufacturer/tradename Axygen Inc AM-2ML-RD-IMP, Corning | manufacturer/tradename Corning, Axygen Inc AM-2ML-SQ-IMP | manufacturer/tradename Corning, Axygen Inc AM-2ML-SQ-IMP-S | manufacturer/tradename Axygen Inc AM-48-IMP, Corning |
| sterility non-sterile | sterility non-sterile | sterility sterile | sterility non-sterile |
| material silicone (research-grade) | material silicone (research-grade) | material silicone (research-grade) | material silicone (research-grade) |
| packaging pkg of 5x10mats/cs | packaging pkg of 5x10mats/cs | packaging pkg of 5x10mats/cs | packaging pkg of 5x10mats/cs |
| size 96 wells | size 96 wells | size 96 wells | size 48 wells |
Still not finding the right product?
Explore all of our products under Impermamat sealing mat, chemical resistant silicone
Scegli una delle versioni più recenti:
Possiedi già questo prodotto?