Vai a
BAF378620002
Scienceware® Hand Held Colony Counter
Includes storage case, red and black pens, and battery
Sinonimo/i:
378620002, F37862-0002, cell counting tool, counting device, counting tool, manual counting tool
Autenticati per visualizzare i prezzi organizzativi e contrattuali.
Scegli un formato
Cambia visualizzazione
Voi/SKUDisponibilitàPrezzo
1 ea
Per conoscere la disponibilità, visualizza il carrello
CHF 439.00
Informazioni su questo articolo
NACRES:
NB.22
UNSPSC Code:
41104000
CHF 439.00
Per conoscere la disponibilità, visualizza il carrello
​
Servizio Tecnico
Hai bisogno di aiuto? Il nostro team di scienziati qualificati è a tua disposizione.
Permettici di aiutarti
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F37862-0002
General description
Provides a cost effective and accurate way to count colonies of bacteria or other microorganisms grown on an agar plate. This hand-held, simple to operate device marks, counts, and gives an audible and visual alert each time a count is registered. The lightweight, pen-style design reduces the likelihood of error, combining all aspects of counting into one motion. CE approved.
Features and Benefits
  • Fast, accurate and economical sample counting and sample count verification for up to 35 dishes
  • Ability to count both up and down to verify sample counts
  • Can display the sum total of all colonies counted
  • Backlit display allows easy viewing in low light conditions
  • Audible alert can be turned off if desired
  • Counting function can be locked to prevent error during breaks between counting
  • Long life, red and black felt-tip markers (included) work on plastic or glass and are oil and water resistant; ink is removable with ethyl alcohol
  • Includes 3V lithium battery and protective storage case
Legal Information
Scienceware is a registered trademark of Bel-Art Products, a division of SP
Confronta articoli simili
Visualizza il confronto per intero
Mostra le differenze
1 of 1
Questo articolo
P7613BAF378633000HS120000
description
Includes storage case, red and black pens, and battery
description
mirror magnification 3X actual plate size
description
Includes Hand-Held Colony Counter (F37862-0002), Mini LED Light Box (F37864-3000) and Mini Magnifier (F37865-0000)
description
-
packaging
pack of 1 ea
packaging
pack of 1 ea
packaging
pack of 1 ea
packaging
-
manufacturer/tradename
Bel-Art F37862-0002
manufacturer/tradename
Bel-Art F37861-0000
manufacturer/tradename
Bel-Art F37863-3000
manufacturer/tradename
Heathrow Scientific HS120000
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)