Scegli un formato
| Voi/SKU | Disponibilità | Prezzo |
|---|
Informazioni su questo articolo
material
stainless steel
packaging
pack of 1 ea
manufacturer/tradename
Benchmark Scientific IPS2050-P-S1
diam. × L
68 mm × 400 mm
compatibility
overhead stirrer equipment
General description
1 of 1
Questo articolo | |||
|---|---|---|---|
| description flat blade, stainless steel | description 4-arm, stainless steel | description centrifugal, PTFE-coated | description flat blade, stainless steel |
| compatibility overhead stirrer equipment | compatibility overhead stirrer equipment | compatibility overhead stirrer equipment | compatibility overhead stirrer equipment |
| material stainless steel | material stainless steel | material PTFE-coated | material stainless steel |
| packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea |
| diam. × L 68 mm × 400 mm | diam. × L 50 mm × 400 mm | diam. × L 85 mm × 350 mm , open | diam. × L 70 mm × 350 mm |
| manufacturer/tradename Benchmark Scientific IPS2050-P-S1 | manufacturer/tradename Benchmark Scientific IPS2050-P-S4 | manufacturer/tradename Benchmark Scientific IPS2050-P-T2 | manufacturer/tradename Benchmark Scientific IPS2050-P-S1 |
Still not finding the right product?
Explore all of our products under Benchmark Overhead Stirrer Propeller
Scegli una delle versioni più recenti:
Possiedi già questo prodotto?