Vai a
EP5804753003
Eppendorf® Adapter For 100 mL Rectangular Bucket
holds standard, Vacutainer, Falcon and micro test tubes 3-15 mL, pack of 2 ea
Autenticati per visualizzare i prezzi organizzativi e contrattuali.
Scegli un formato
Cambia visualizzazione
Voi/SKUDisponibilitàPrezzo
2 ea
Per conoscere la disponibilità, visualizza il carrello
CHF 176.00
Informazioni su questo articolo
NACRES:
NB.82
UNSPSC Code:
41103911
Compatibility:
for use with 100 mL rectangular bucket
Manufacturer/tradename:
Eppendorf® 5804753003
CHF 176.00
Per conoscere la disponibilità, visualizza il carrello
​
Servizio Tecnico
Hai bisogno di aiuto? Il nostro team di scienziati qualificati è a tua disposizione.
Permettici di aiutarti
feature
holds standard, Vacutainer, Falcon and micro test tubes 3-15 mL
packaging
pack of 2 ea
manufacturer/tradename
Eppendorf® 5804753003
technique(s)
centrifugation: suitable
compatibility
for use with 100 mL rectangular bucket
General description
Tube adapters for use with Eppendorf 100 mL rectangular bucket.
Legal Information
Eppendorf is a registered trademark of Eppendorf SE
Confronta articoli simili
Visualizza il confronto per intero
Mostra le differenze
1 of 1
Questo articolo
EP5804755006EP5804761006EP5804718003
description
holds standard, Vacutainer, Falcon and micro test tubes 3-15 mL, pack of 2 ea
description
holds standard, Vacutainer, Falcon and micro test tubes 15 mL Falcon, pack of 2 ea
description
holds 50 mL Falcon tubes, pack of 2 ea
description
holds standard, Vacutainer, Falcon and micro test tubes 50 mL Falcon, pack of 2 ea
manufacturer/tradename
Eppendorf® 5804753003
manufacturer/tradename
Eppendorf® 5804755006
manufacturer/tradename
Eppendorf® 5804761006
manufacturer/tradename
Eppendorf® 5804718003
compatibility
for use with 100 mL rectangular bucket
compatibility
for use with 100 mL rectangular bucket
compatibility
for use with 100 mL rectangular bucket
compatibility
for use with 100 mL rectangular bucket
packaging
pack of 2 ea
packaging
pack of 2 ea
packaging
pack of 2 ea
packaging
pack of 2 ea
technique(s)
centrifugation: suitable
technique(s)
centrifugation: suitable
technique(s)
centrifugation: suitable
technique(s)
centrifugation: suitable
feature
holds standard, Vacutainer, Falcon and micro test tubes 3-15 mL
feature
holds standard, Vacutainer, Falcon and micro test tubes 15 mL Falcon
feature
holds 50 mL Falcon tubes
feature
holds standard, Vacutainer, Falcon and micro test tubes 50 mL Falcon
Still not finding the right product?
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)