Vai a
EP5820767006
Spare Sealing
pack of 5
Sinonimo/i:
Lid for Eppendorf® FA-45-48-11 Rotor
Autenticati per visualizzare i prezzi organizzativi e contrattuali.
Scegli un formato
Cambia visualizzazione
Voi/SKUDisponibilitàPrezzo
5 ea
Per conoscere la disponibilità, visualizza il carrello
CHF 230.00
Informazioni su questo articolo
NACRES:
NB.82
UNSPSC Code:
41103900
Packaging:
pack of 5
CHF 230.00
Per conoscere la disponibilità, visualizza il carrello
​
Servizio Tecnico
Hai bisogno di aiuto? Il nostro team di scienziati qualificati è a tua disposizione.
Permettici di aiutarti
feature
For rotor FA-45-24-11-Kit, FA-45-20-17, FA-30x2 and FA-48x2. Also for rotors FA-45-48-11 manufactured after 05/2019
packaging
pack of 5
manufacturer/tradename
Eppendorf® 5820767006 (Global)
General description
Eppendorf® is a top company in the field of life science. They create and sell tools, materials, and services for handling liquids, samples, and cells in laboratories around the world. Their products are widely used in research labs at universities and companies. The spare sealing is suitable for use with an Eppendorf® FA-45-48-11 Rotor lid.
Legal Information
Eppendorf is a registered trademark of Eppendorf SE
Confronta articoli simili
Visualizza il confronto per intero
Mostra le differenze
1 of 1
Questo articolo
EP5820762004EP5409719009EP5820749008
Spare Sealing pack of 5
EP5820767006
Spare Sealing
Spare Sealing for Eppendorf® FA-45-30-11 rotor lid, pack of 5
EP5820762004
Spare Sealing
Spare Sealing for Eppendorf® S-24-11-AT rotor lid, pack of 5
EP5409719009
Spare Sealing
Spare Sealing for Eppendorf® for S-4-104 rotor lid, for 750 mL round buckets, pack of 5
EP5820749008
Spare Sealing
description
pack of 5
description
for Eppendorf® FA-45-30-11 rotor lid, pack of 5
description
for Eppendorf® S-24-11-AT rotor lid, pack of 5
description
for Eppendorf® for S-4-104 rotor lid, for 750 mL round buckets, pack of 5
packaging
pack of 5
packaging
pack of 5
packaging
pack of 5
packaging
pack of 5
feature
For rotor FA-45-24-11-Kit, FA-45-20-17, FA-30x2 and FA-48x2. Also for rotors FA-45-48-11 manufactured after 05/2019
feature
-
feature
for Eppendorf® S-24-11-AT rotor lid
feature
for Rotors S-4-104, S-4×750, S-4×1000, Round cups 750 mL/1000 mL
manufacturer/tradename
Eppendorf® 5820767006 (Global)
manufacturer/tradename
Eppendorf® 5820762004 (Global)
manufacturer/tradename
Eppendorf® 5409719009 (Global)
manufacturer/tradename
Eppendorf® 5820749008 (Global)
Still not finding the right product?
Explore all of our products under Spare Sealing
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)