Vai a
MTCF4062F
MTC™ Bio BioFlask™ Erlenmeyer Shaker Flask, Individually Wrapped
capacity 500 mL, polypropylene, flat bottom, cap, vented, sterile
Sinonimo/i:
culture flask, MTC Bio F4062-F, laboratory flask, plastic flask, shaker flask
Autenticati per visualizzare i prezzi organizzativi e contrattuali.
Scegli un formato
Cambia visualizzazione
Voi/SKUDisponibilitàPrezzo
Informazioni su questo articolo
NACRES:
NB.15
UNSPSC Code:
41122104
Capacity:
500 mL
Material:
flat bottom, polypropylene
Sterility:
sterile
Feature:
cap vented
Prezzi e disponibilità al momento non sono disponibili
Servizio Tecnico
Hai bisogno di aiuto? Il nostro team di scienziati qualificati è a tua disposizione.
Permettici di aiutarti
material
flat bottom, polypropylene
sterility
sterile
feature
cap vented
packaging
pack of 12 ea (individually wrapped)
manufacturer/tradename
MTC™ Bio F4062-F
capacity
500 mL
General description
Erlenmeyer Shaker Flasks are made of polycarbonate or PETG. The polypropylene screw caps are vented with 0.2µm membranes to increase air transmission and enhance cell growth. The mouth size conforms with the international SBS standard, for quick, secure connection to bioreactor equipment. Polycarbonate flasks have baffled (ribbed) bottoms to provide increased oxygenation and mixing efficiency even at lower shaking speeds. Polycarbonate Flasks are autoclavable at 121°C and 15 psi. All flasks, regardless of size, are individually wrapped and gamma sterilized to SAL 10-6. They are free of RNase, DNase, pyrogen, endotoxins and human DNA.
Flask, Erlenmeyer, 500ml, PETG, flat bottom w / vented screw cap, Sterile, indiv wrapped, 12/CS
Features and Benefits
  • Vented screw caps
  • Flat or baffled bottoms
  • Gamma sterile (SAL 10-6)
  • Pyrogen free and RNase/DNase free
  • Individually wrapped
Legal Information
BioFlask is a trademark of MTC Bio
MTC is a trademark of MTC Bio
Confronta articoli simili
Visualizza il confronto per intero
Mostra le differenze
1 of 1
Questo articolo
MTCF4062BMTCF4063FMTCF4061B
description
capacity 500 mL, polypropylene, flat bottom, cap, vented, sterile
description
capacity 500 mL, polycarbonate, bottom (baffled), cap, vented, sterile
description
capacity 1000 mL, polypropylene, flat bottom, cap (vented), sterile
description
capacity 250 mL, polycarbonate, bottom (baffled), cap, vented, sterile
manufacturer/tradename
MTC™ Bio F4062-F
manufacturer/tradename
MTC™ Bio F4062-B
manufacturer/tradename
MTC™ Bio F4063-F
manufacturer/tradename
MTC™ Bio F4061-B
capacity
500 mL
capacity
500 mL
capacity
1000 mL
capacity
250 mL
sterility
sterile
sterility
sterile
sterility
sterile
sterility
sterile
material
polypropylene , flat bottom
material
bottom (baffled), polycarbonate
material
flat bottom, polypropylene , cap (vented)
material
bottom (baffled), polycarbonate
feature
cap vented
feature
cap vented
feature
-
feature
cap vented
packaging
pack of 12 ea (individually wrapped)
packaging
pack of 12 ea (individually wrapped)
packaging
pack of 6 ea (individually wrapped)
packaging
pack of 12 ea (individually wrapped)
Still not finding the right product?
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)