O této položce
UNSPSC Code:
41113036
eCl@ss:
32040303
Material:
polystyrene
Sterility:
sterile
Binding type:
poly-D -lysine coated surface
Feature:
lid, wells 384
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohlimaterial
polystyrene
sterility
sterile
feature
lid, wells 384
packaging
case of 100 (5 packs of 20)
manufacturer/tradename
Corning 3769
plate wells
95 μL
plate working volume
20 μL (80 μL)
binding type
poly-
General description
- 20 per Bag, Lid, Aseptically Manufactured
- White walled plates enhance luminescent signals and have low background luminescence and fluorescence
- Opaque walls to prevent well-to-well crosstalk
- Optically clear flat well bottom permits direct microscopic viewing
- Can be used for both top and bottom reading instruments
- Flat bottoms with 112 µL total volume
- Recommended working volumes of 20 to 80 µL
- Poly-D-lysine coated for enhanced cell attachment
- Aseptically manufactured and nonpyrogenic Individual alphanumeric codes for well identification
- Bulk packed 20 per bag
Application
assay plate
Legal Information
Corning is a registered trademark of Corning, Inc.
Still not finding the right product?
Explore all of our products under Corning® Microplate
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?