Přihlásit pro zobrazení organizačních a smluvních cen.
Vybrat velikost
Změnit zobrazení
| Vám/SKU | Dostupnost | Cena |
|---|
O této položce
UNSPSC Code:
12352204
Informace o stanovených cenách a dostupnosti nejsou v současnosti dostupné.
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohliGeneral description
High Temp M-MLV (Moloney Murine Leukemia Virus) Reverse Transcriptase (HTRT) DNA polymerase uses DNA, single-stranded RNA, or an RNA:DNA hybrid (using a primer) to synthesize a complementary DNA strand . Wild type (WT) and traditional (MMLV-RT) are restricted to lower temperatures (up to 50℃) during cDNA synthesis. For many reverse transcription reactions, temperatures higher than 50℃ are beneficial. Higher temperatures reduce the chances of inhibition due to RNA secondary structures, while at the same time increasing primer binding specificity. These benefits lead to an increase in desired cDNA yield from the reverse transcription reaction. HTRT maintains its performance at high temperatures and outcompetes WT MMLV-RTs. This enzyme is isolated from E. coli expressing a portion of the pol gene of M-MLV on a plasmid.
Application
HTRT reverse transcriptase is used for:
- preparation of cDNA libraries
- first strand cDNA synthesis for use in RT-PCR
- 2-step RT-PCR assay
- Quantitative realtime-polymerase chain reaction (RT-qPCR)
1 of 1
Tato položka | |||
|---|---|---|---|
| description Thermostable Moloney Murine Leukemia Virus enzyme & buffer for cDNA synthesis | description Moloney Murine Leukemia Virus enzyme & buffer for cDNA synthesis | description pkg of 250 x 25 µL reactions, active at high temperatures, lyo-compatible, for fast synthesis of short cDNA (< 1 Kb) | description For reverse transcription at higher temperatures & rare mRNAs |
| form liquid | form liquid | form - | form crystalline powder |
| shipped in dry ice | shipped in wet ice | shipped in dry ice | shipped in wet ice |
| storage condition OK to freeze | storage condition - | storage condition - | storage condition - |
| Quality Level 200 | Quality Level 200 | Quality Level - | Quality Level 200 |
WGK
WGK 3
Skladovací třída
10 - Combustible liquids
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?
Dokumenty související s produkty, které jste v minulosti zakoupili, byly za účelem usnadnění shromážděny ve vaší Knihovně dokumentů.



![Enhanced Avian Reverse Transcriptase [eAMV™ RT] For reverse transcription at higher temperatures & rare mRNAs](/deepweb/assets/sigmaaldrich/product/images/496/245/af9bcef6-1474-494b-8fba-32ca0fd56c42/640/af9bcef6-1474-494b-8fba-32ca0fd56c42.jpg)