Přihlásit pro zobrazení organizačních a smluvních cen.
Vybrat velikost
Změnit zobrazení
O této položce
UNSPSC Code:
41116104
eCl@ss:
40050204
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohliusage
sufficient for 100 colorimetric tests
Quality Segment
application(s)
environmental
food and beverages
life science and biopharma
detection method
colorimetric
shipped in
dry ice
storage temp.
−20°C
General description
Acetate is the ion of acetic acid and plays a major part in human metabolism. Acetate is generated through the metabolism of carbohydrates, amino acids, fatty acids, and ethanol. It serves as a crucial precursor for multiple biochemical pathways, including the biosynthesis of acetylcholine, neurotransmitters, and the acetylation of proteins, which is vital for gene regulation and protein function. Acetate and the related metabolism of acetyl-CoA confers numerous metabolic functions, including energy production, lipid synthesis and protein acetylation.
Biochem/physiol Actions
The Acetate Colorimetric Assay Kit employs a coupled enzyme reaction to quantify acetate levels in various biological and food samples. This assay kit operates on a principle where acetate reacts with specific enzymes, resulting in a color change that can be measured spectrophotometrically at 450 nm. This colorimetric measurement is directly proportional to the acetate concentration in the sample, providing a straightforward and accurate method for acetate quantification.
Features and Benefits
Compatibility with High-Throughput Systems: Seamlessly integrate our kit into high-throughput handling systems, ensuring smooth operations and accurate results, enhancing efficiency in your laboratory workflow.
Convenient Data Analysis: Supportive calculator will calculate the reagents needed according to your experimental needs, and analyze your results based on your experimental data.
Click here to download a calculator excel file
Convenient Data Analysis: Supportive calculator will calculate the reagents needed according to your experimental needs, and analyze your results based on your experimental data.
Click here to download a calculator excel file
Still not finding the right product?
Explore all of our products under Acetate Assay Kit
Výstražné slovo
Danger
Kódy nebezpečí
Hazard Classifications
Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1
Skladovací třída
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Vyberte jednu z posledních verzí:
Již tento produkt vlastníte?
Dokumenty související s produkty, které jste v minulosti zakoupili, byly za účelem usnadnění shromážděny ve vaší Knihovně dokumentů.

