Přeskočit na
R1525
Texan™ reagent reservoir for multichannel pipettes
with lid, non-sterile, pack of 100 ea (10 pavks of 10ea)
Synonyma:
multichannel pipette reservoir, reagent reservoir
Přihlásit pro zobrazení organizačních a smluvních cen.
Vybrat velikost
Změnit zobrazení
Vám/SKUDostupnostCena
100 ea
Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti.
6 470,00 Kč
O této položce
NACRES:
NB.01
UNSPSC Code:
41121522
eCl@ss:
32070102
Manufacturer/tradename:
Excel Scientific RSV-L175-N10
Capacity:
175 mL
Material:
V-bottom bottom, colorless , polypropylene
Sterility:
non-sterile
6 470,00 Kč
Obraťte se na zákaznický servis a vyžádejte si informaci o dostupnosti.
​
Technický servis
Potřebujete pomoc? Náš tým zkušených odborníků je vám k dispozici.
Dovolte nám, abychom vám pomohli
material
V-bottom bottom, colorless , polypropylene
sterility
non-sterile
packaging
pack of 100 ea (10 pavks of 10ea)
manufacturer/tradename
Excel Scientific RSV-L175-N10
capacity
175 mL
General description
Texan reagent reservoirs are reusable -- an environmentally sound alternative to disposable reagent reservoirs. Their extra-large capacity means fewer refills and time saved. The V-shaped basin is compatible with both eight- and twelve-place multiple pipettors with minimal reagent waste. Lids are removable -- attached with ball and socket hinges -- and latch tightly for secure storage of left-over reagents. With lids closed, Texan reagent reservoirs are stackable. A perimeter ridge on the lid prevents sliding. Construction is clear, chemical-resistant, autoclavable polypropylene. Molded graduation marks provide volume indication at 25, 50, 75, 100, 125, and 150 mL.
Legal Information
Texan is a trademark of Excel Scientific, Inc.
Porovnat podobné položky
Zobrazit úplné porovnání
Zobrazit rozdíly
1 of 1
Tato položka
R9259BR703411BR703459
description
with lid, non-sterile, pack of 100 ea (10 pavks of 10ea)
description
without lid, non-sterile, pack of 100 ea
description
capacity 60 mL, pack of 100 ea, without lid
description
capacity 60 mL
manufacturer/tradename
Excel Scientific RSV-L175-N10
manufacturer/tradename
Excel Scientific RSV-0175-N10
manufacturer/tradename
BRAND 703411
manufacturer/tradename
BRAND 703459
capacity
175 mL
capacity
175 mL
capacity
60 mL
capacity
60 mL
sterility
non-sterile
sterility
non-sterile
sterility
sterile
sterility
non-sterile
material
V-bottom bottom, polypropylene , colorless
material
colorless , V-bottom bottom, polypropylene
material
V-bottom bottom, colorless , polypropylene
material
polypropylene , colorless , V-bottom bottom
packaging
pack of 100 ea (10 pavks of 10ea)
packaging
pack of 100 ea
packaging
pack of 100 ea
packaging
pack of 10 ea
Still not finding the right product?
Vyberte jednu z posledních verzí:
Osvědčení o analýze (COA)