Skip to Content
Merck

EMU080871

MISSION® esiRNA

targeting mouse Capn2

Sign In to View Organizational & Contract Pricing.

Select a Size

20 μG

CZK 5,620.00

50 μG

CZK 10,100.00

CZK 5,620.00


Please contact Customer Service for Availability


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CCAAGCTGTTGATGACGAGATCGAGGCCAACATTGAAGAGATTGACGCCAATGAGGAGGACATTGACGATGGATTCCGAAGGCTCTTTGTTCAGCTGGCTGGAGAGGATGCAGAGATCTCTGCCTTTGAGCTGCAGACCATCTTAAGAAGAGTTCTAGCCAAACGCCAAGACATCAAGTCAGACGGCTTCAGCATCGAGACCTGTAAGATCATGGTGGACATGCTGGATGAAGATGGGAGTGGCAAGCTTGGCCTGAAGGAGTTCTACATCCTCTGGACGAAGATTCAGAAATACCAAAAAATCTACCGGGAAATCGATGTGGACAGGTCTGGAACCATGAATTCCTACGAGATGCGGAAAGCACTGGAAGAAGCAGGTTTCAAGCTGCCCTGTCAACTTCATCAAGTCATCGTTGCCCGGTTTGCAGACGACGAGCTAATCATCGACTTTGACAATTTTGTGCGGTGTT

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
EMU057001EMU184601EMU071211
Gene Information

mouse ... CAPN2(12334), Capn2(12334)

Gene Information

mouse ... CAPN1(12333), Capn1(12333)

Gene Information

mouse ... CAPNS2(69543), Capns2(69543)

Gene Information

mouse ... CAPG(12332), Capg(12332)

product line

MISSION®

product line

MISSION®

product line

MISSION®

product line

MISSION®

esiRNA cDNA target sequence

CCAAGCTGTTGATGACGAGATCGAGGCCAACATTGAAGAGATTGACGCCAATGAGGAGGACATTGACGATGGATTCCGAAGGCTCTTTGTTCAGCTGGCTGGAGAGGATGCAGAGATCTCTGCCTTTGAGCTGCAGACCATCTTAAGAAGAGTTCTAGCCAAACGCCAAGACATCAAGTCAGACGGCTTCAGCATCGAGACCTGTAAGATCATGGTGGACATGCTGGATGAAGATGGGAGTGGCAAGCTTGGCCTGAAGGAGTTCTACATCCTCTGGACGAAGATTCAGAAATACCAAAAAATCTACCGGGAAATCGATGTGGACAGGTCTGGAACCATGAATTCCTACGAGATGCGGAAAGCACTGGAAGAAGCAGGTTTCAAGCTGCCCTGTCAACTTCATCAAGTCATCGTTGCCCGGTTTGCAGACGACGAGCTAATCATCGACTTTGACAATTTTGTGCGGTGTT

esiRNA cDNA target sequence

CCAACCTCCCTGATGAGAAAGTTCTCTCTGAAGAGGAGATTGATGACAACTTCAAGACCTTGTTCAGCAAGTTGGCAGGGGATGACATGGAGATCAGCGTCAAGGAGCTACAGACCATCTTGAACAGGATCATTAGCAAACACAAAGACCTGCGCACTAATGGCTTCAGCCTGGAGTCGTGCCGCAGCATGGTGAACCTCATGGATCGAGATGGCAACGGGAAGCTGGGTCTGGTGGAGTTCAACATCCTGTGGAACCGCATCCGAAATTACCTGACCATCTTCCGGAAGTTTGACCTGGACAAGTCTGGCAGCATGAGTGCCTATGAGATGAGGATGGCCATCGAGGCTGCAGGCTTCAAGCTTAACAAGAAGCTGCATGAACTCATAATCACCCGCTACTCGGAGCCCGACCTGGCCGTGGACTTTGACAACTTTGTGTGCTGTCTTGTGCGGCTGGAGACCATGTTCCGGTTTTTCAAACTTCTGGACACAGACCTGGATGGTGTTG

esiRNA cDNA target sequence

AATCCTTGGAGGAATTGTCAATTTTATCAGCGAGGCTGCCGCAGCTCAGTACACCCCAGAGCCGCCTCCCCAACAGCAGCATTTTACCGTCGTGGAGGCCTCAGAAAGTGAGGAAGTCAGACGTTTTCGGCAGCAATTTACACAGCTGGCTGGACCCGACATGGAGGTGGGTGCTACTGACCTGATGAATATTCTCAACAAAGTCCTTTCTAAGCATAAAGAGCTGAAGACAGAGGGCTTCAGCCTTGATACCTGTAGGAGCATTGTGTCTGTCATGGACAGTGACACCACAGGGAAACTGGGATTTGAGGAATTTAAGTACCTGTGGAACAACATCAAGAAATGGCAGTGTGTTTTCAAGCAGTACGACAGTGACCATTCTGGCTCCCTCGGAAGCTCCCAGCTGCACGGGGCCATGCAGGCAGCGGGCTTCCAGCTCAATGAGCAACTTTACCTAATGATTGTCCGTCGGTATGCTGACGAAGATGGCGGGATGGATTTTAACAACTTTATCAGCTGC

esiRNA cDNA target sequence

GTGGTGGTAGAGTCGGCATTTCACAAGACAACCTTCGCGCCACCCCAGCAGCCATCAGGAAGCTCTACCAGGTTAAGGGGAAGAAGAACATCCGTGCGACCGAGAGGGCTCTGAGTTGGGACAGCTTCAACACTGGGGACTGCTTCATCCTGGACCTGGGTCAGAACATCTTTGCCTGGTGTGGTGGAAAGTCCAACATCCTTGAGCGCAACAAGGCGAGGGACCTGGCCCTGGCCATCAGGGACAGCGAGCGGCAGGGCAAGGCCCAGGTGGAAATCATCACTGATGGAGAGGAGCCAGCCGAGATGATTCAGGTTCTGGGCCCCAAGCCTGCTCTGAAGGAGGGTAACCCCGAGGAAGACATTACAGCTGACCAGACCAACGCCCAGGCTGCAGCCCTGTATAAGGTCTCTGATGCCACTGGACAG

Ensembl | mouse accession no.

ENSMUSG00000026509

Ensembl | mouse accession no.

ENSMUSG00000024942

Ensembl | mouse accession no.

ENSMUSG00000078144

Ensembl | mouse accession no.

ENSMUSG00000056737

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service