Merck
  • Genetic analysis of Hedgehog signaling in ventral body wall development and the onset of omphalocele formation.
Genetic analysis of Hedgehog signaling in ventral body wall development and the onset of omphalocele formation.
PloS one (2011-02-02)
Daisuke Matsumaru, Ryuma Haraguchi, Shinichi Miyagawa, Jun Motoyama, Naomi Nakagata, Frits Meijlink, Gen Yamada
ABSTRACT
An omphalocele is one of the major ventral body wall malformations and is characterized by abnormally herniated viscera from the body trunk. It has been frequently found to be associated with other structural malformations, such as genitourinary malformations and digit abnormalities. In spite of its clinical importance, the etiology of omphalocele formation is still controversial. Hedgehog (Hh) signaling is one of the essential growth factor signaling pathways involved in the formation of the limbs and urogenital system. However, the relationship between Hh signaling and ventral body wall formation remains unclear. To gain insight into the roles of Hh signaling in ventral body wall formation and its malformation, we analyzed phenotypes of mouse mutants of Sonic hedgehog (Shh), GLI-Kruppel family member 3 (Gli3) and Aristaless-like homeobox 4 (Alx4). Introduction of additional Alx4(Lst) mutations into the Gli3(Xt/Xt) background resulted in various degrees of severe omphalocele and pubic diastasis. In addition, loss of a single Shh allele restored the omphalocele and pubic symphysis of Gli3(Xt/+); Alx4(Lst/Lst) embryos. We also observed ectopic Hh activity in the ventral body wall region of Gli3(Xt/Xt) embryos. Moreover, tamoxifen-inducible gain-of-function experiments to induce ectopic Hh signaling revealed Hh signal dose-dependent formation of omphaloceles. We suggest that one of the possible causes of omphalocele and pubic diastasis is ectopically-induced Hh signaling. To our knowledge, this would be the first demonstration of the involvement of Hh signaling in ventral body wall malformation and the genetic rescue of omphalocele phenotypes.
Sigma-Aldrich
Alizarin Red S, certified by the BSC
Sigma-Aldrich
Alcian Blue 8GX, powder or crystals, certified by the BSC
Sigma-Aldrich
Alcian Blue 8GX, powder
Sigma-Aldrich
Alcian Blue 8GX
Sigma-Aldrich
Alcian Blue 8GX, BioReagent, suitable for detection of glycoproteins
Sigma-Aldrich
Acridine Orange base, Dye content 75 %
Sigma-Aldrich
Paraformaldehyde, prilled, 95%
Sigma-Aldrich
Paraformaldehyde, meets analytical specification of DAC, 95.0-100.5%
Sigma-Aldrich
Paraformaldehyde, reagent grade, crystalline
Sigma-Aldrich
Paraformaldehyde, powder, 95%