Merck
  • Conformational change in transfer RNA is an early indicator of acute cellular damage.
Conformational change in transfer RNA is an early indicator of acute cellular damage.
Journal of the American Society of Nephrology : JASN (2014-05-17)
Eikan Mishima, Chisako Inoue, Daisuke Saigusa, Ryusuke Inoue, Koki Ito, Yusuke Suzuki, Daisuke Jinno, Yuri Tsukui, Yosuke Akamatsu, Masatake Araki, Kimi Araki, Ritsuko Shimizu, Haruka Shinke, Takehiro Suzuki, Yoichi Takeuchi, Hisato Shima, Yasutoshi Akiyama, Takafumi Toyohara, Chitose Suzuki, Yoshikatu Saiki, Teiji Tominaga, Shigehito Miyagi, Naoki Kawagisihi, Tomoyoshi Soga, Takayoshi Ohkubo, Kenichi Yamamura, Yutaka Imai, Satohiro Masuda, Venkata Sabbisetti, Takaharu Ichimura, David B Mount, Joseph V Bonventre, Sadayoshi Ito, Yoshihisa Tomioka, Kunihiko Itoh, Takaaki Abe
ABSTRACT
Tissue damage by oxidative stress is a key pathogenic mechanism in various diseases, including AKI and CKD. Thus, early detection of oxidative tissue damage is important. Using a tRNA-specific modified nucleoside 1-methyladenosine (m1A) antibody, we show that oxidative stress induces a direct conformational change in tRNA structure that promotes subsequent tRNA fragmentation and occurs much earlier than DNA damage. In various models of tissue damage (ischemic reperfusion, toxic injury, and irradiation), the levels of circulating tRNA derivatives increased rapidly. In humans, the levels of circulating tRNA derivatives also increased under conditions of acute renal ischemia, even before levels of other known tissue damage markers increased. Notably, the level of circulating free m1A correlated with mortality in the general population (n=1033) over a mean follow-up of 6.7 years. Compared with healthy controls, patients with CKD had higher levels of circulating free m1A, which were reduced by treatment with pitavastatin (2 mg/d; n=29). Therefore, tRNA damage reflects early oxidative stress damage, and detection of tRNA damage may be a useful tool for identifying organ damage and forming a clinical prognosis.
Sigma-Aldrich
Ribonucleic acid, transfer from baker′s yeast (S. cerevisiae), buffered aqueous solution
Sigma-Aldrich
Ribonucleic acid, transfer from baker′s yeast (S. cerevisiae), buffered aqueous solution
Sigma-Aldrich
Ribonucleic acid, transfer from baker′s yeast (S. cerevisiae), Type X-SA, lyophilized powder
Sigma-Aldrich
Ribonucleic acid, transfer from bovine liver, Type XI, lyophilized powder
Sigma-Aldrich
Ribonucleic acid, transfer from wheat germ, Type V, 15-19 units/mg solid
Sigma-Aldrich
Adenosine
Sigma-Aldrich
Adenosine, BioReagent, suitable for cell culture
Sigma-Aldrich
Adenosine, ≥99%
Sigma-Aldrich
Ribonucleic acid, transfer from Escherichia coli, Type XX, Strain W, lyophilized powder
Adenosine, European Pharmacopoeia (EP) Reference Standard
Supelco
Adenosine, Pharmaceutical Secondary Standard; Certified Reference Material