Merck
  • Poly(ADP-Ribose) Polymerase Inhibition Improves Corneal Epithelial Innervation and Wound Healing in Diabetic Rats.
Poly(ADP-Ribose) Polymerase Inhibition Improves Corneal Epithelial Innervation and Wound Healing in Diabetic Rats.
Investigative ophthalmology & visual science (2015-02-26)
Yong-Soo Byun, Borami Kang, Young-Sik Yoo, Choun-Ki Joo
ABSTRACT
We evaluated the effect of poly(ADP-ribose) polymerase (PARP) inhibition by using 1,5-isoquinolinediol (ISO) on corneal epithelial innervation in diabetic rats. ISO (3 mg/kg, intraperitoneal) or vehicle was administered to rats with diabetes induced by streptozotocin for 4 weeks. Epithelial innervation, epithelial wound healing, and corneal sensation were evaluated in diabetic rats (DM rats), diabetic rats treated with ISO (DM-ISO rats), and nondiabetic (non-DM) rats. The density of epithelial innervation was calculated separately as nerve terminals and sub-basal nerve plexus by analyzing the images of whole-mount corneas. Healed areas of epithelial defect were measured at 0, 18, and 36 hours after creating a 4-mm wound on the cornea. Corneal sensitivity test was conducted using a Cochet-Bonnet handheld esthesiometer. Additionally, PARP1 and poly(ADP-ribosyl)ated polymers (pADPr) as its products, were identified in trigeminal ganglions (TGs) by Western blot analysis and immunofluorescence staining. In DM rats, the density of nerve terminals (5.57% ± 0.94%) and sub-basal nerve plexus (22.08 ± 1.78 mm/mm(2)) was significantly reduced in comparison with that in DM-ISO rats (8.64% ± 1.42%, 30.82 ± 2.01 mm/mm(2), respectively) and non-DM rats (9.02 ± 1.14%, 34.77 ± 4.45 mm/mm(2), respectively). The percentages of healed area of the epithelial defects at 18 and 36 hours were significantly smaller in DM rats (23.8 ± 5.2%, 53.2 ± 4.6%, respectively) than in DM-ISO rats (43.2 ± 1.4%, 75.8 ± 2.2%, respectively) and non-DM rats (48.1 ± 8.6%, 86.1 ± 3.3%, respectively). Corneal sensitivity decreased in DM rats (51.1 ± 0.3 mm) but not in DM-ISO rats (57.8 ± 0.2 mm). There were no differences between parameters in DM-ISO rats and those in non-DM rats. Diabetic corneas showed loss of epithelial innervation, resulting in delayed epithelial healing and decreased corneal sensitivity. Inhibition of poly(ADP-ribose) polymerase (PARP) with 1,5-isoquinolinediol alleviated these diabetes-induced alterations in the corneal epithelium in the diabetic rats.
Product Description
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
USP
Lidocaine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
USP
Levofloxacin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium chloride solution, 0.85%
Supelco
Lidocaine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Lidocaine, powder
Sigma-Aldrich
Lidocaine, analytical standard
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Xylazine hydrochloride, ≥99.0% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)