Merck
  • The synergistic effect of ultrasound and chemical penetration enhancers on chorioamnion mass transport.
The synergistic effect of ultrasound and chemical penetration enhancers on chorioamnion mass transport.
Journal of controlled release : official journal of the Controlled Release Society (2014-12-30)
Aharon Azagury, Luai Khoury, Yair Adato, Lior Wolloch, Ilana Ariel, Mordechai Hallak, Joseph Kost
ABSTRACT
In our previous study we proposed the use of chemical penetration enhancers for noninvasive detection of fetus abnormalities that can also be utilized for direct fetal drug delivery. In an attempt to further increase the mass transport rate across the amniotic membrane, thus shortening the procedure and improving the applicability of the proposed procedure, the effect and mechanism of combining ultrasound exposure with chemical penetration enhancers' application were assessed. The combined effect was evaluated in vitro on post-delivery human amniotic membrane and ex vivo on rat's whole amniotic sac. Ultrasound effect has been assessed by dye experiments using a customized image analysis program. Additional insights of ultrasound effect's mechanism on biological membranes are presented. Previously we have determined that chemical penetration enhancers affect the fetal membranes via two mechanisms termed as 'extractors' and 'fluidizers'. In this study, we found that combining ultrasound with a 'fluidizer' CPE (e.g. bupivacaine) results in a synergistic enhancement (90-fold) of fetal membrane's mass transport, while combining ultrasound with 'extractors' (e.g. ethanol and NMP) results in an antagonistic effect. The combined procedure is faster and gain greater accuracy than the applications of sole chemical penetration enhancers.
Sigma-Aldrich
Potassium hydride, 30 wt % dispersion in mineral oil
Sigma-Aldrich
Fluorescein, free acid
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Indigo, synthetic, Dye content 95 %
Sigma-Aldrich
Potassium hydride, in paraffin
Sigma-Aldrich
Potassium, chunks (in mineral oil), 98% trace metals basis
Fluorescein, European Pharmacopoeia (EP) Reference Standard
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Supelco
1-Methyl-2-pyrrolidinone, analytical standard
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Lidocaine, analytical standard
Sigma-Aldrich
Lidocaine, powder
Sigma-Aldrich
Xylazine, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Lidocaine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
1-Methyl-2-pyrrolidinone, ReagentPlus®, 99%
USP
Lidocaine, United States Pharmacopeia (USP) Reference Standard