Merck
  • Aptamer-conjugated magnetic nanoparticles for extraction of adenosine from urine followed by electrospray ion mobility spectrometry.
Aptamer-conjugated magnetic nanoparticles for extraction of adenosine from urine followed by electrospray ion mobility spectrometry.
Journal of pharmaceutical and biomedical analysis (2015-01-28)
Marzieh Enteshari Najafabadi, Taghi Khayamian, Zahra Hashemian
ABSTRACT
Magnetic nanoparticles (MNPs) conjugated with aptamer was developed for the selective extraction of adenosine in urine samples followed by electrospray ionization-ion mobility spectrometry (ESI-IMS). The ion mobility spectrum of adenosine showed two peaks at low concentrations and two more peaks related to dimer of adenosine at high concentrations. However, the ion mobility spectrum of eluent at low concentration showed only the peaks related to dimer of adenosine. In other words, aptamer captured two adenosine molecules between the top G-quartet and the two short stems, where they bonded to each other. The mass spectrum of the eluent also validated the presence of dimer (m/z 535.95). The effect of extraction parameters on extraction efficiency including sorbent amount, elution conditions (solvent type and volume) and adsorption conditions were investigated. Under the optimized conditions, the linear dynamic range was found to be 0.05-5.00 μg mL(-1) with detection limit of 0.02 μg mL(-1). The extraction efficiency was 94% and the relative standard deviation was 4% for three replicate measurements of adenosine at 0.25 μg mL(-1) in urine samples. As a practical application, the method was applied for the determination of adenosine in urine samples of patients with lung cancer, and the obtained results were in good agreement with those obtained by HPLC-UV method. Therefore, the proposed method is an alternative clinical analysis.
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, packaged for use in deposition systems, ≥98%
Millipore
Bifido Selective Supplement B, suitable for microbiology
Supelco
Adenosine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
5α-Androstan-17β-ol-3-one, VETRANAL®, analytical standard
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Ammonium hydroxide solution, BioUltra, ~1 M NH3 in H2O (T)
Sigma-Aldrich
Adenosine
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, 99%
Sigma-Aldrich
Adenosine, ≥99%
Sigma-Aldrich
Adenosine, BioReagent, suitable for cell culture
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98%
Adenosine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98.0%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Ammonium hydroxide solution, ACS reagent, 28.0-30.0% NH3 basis
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Ammonium hydroxide solution, 28% NH3 in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethoxysilane, 95%
Supelco
Acetic acid, analytical standard