Skip to Content
Merck

810348P

Avanti

meso-Cl Cyanine 7.5 free acid

new

Avanti Research - A Croda Brand

Synonym(s):

3-(5-carboxypentyl)-2-((E)-2-((E)-2-chloro-3-((E)-2-(1,1,3-trimethyl-1,3-dihydro-2H-benzo[e]indol-2-ylidene)ethylidene)cyclohex-1-en-1-yl)vinyl)-1,1-dimethyl-1H-benzo[e]indol-3-ium chloride, meso-Cl Cyanine 7.5 free acid

Sign In to View Organizational & Contract Pricing

Select a Size

Pricing and availability is not currently available.

About This Item

Empirical Formula (Hill Notation):
C45H48Cl2N2O2
Molecular Weight:
719.78
UNSPSC Code:
12352214

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 5 mg 810348P-5MG

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

fluorescent lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[Cl-].CN1\C(=C\C=C2/CCCC(\C=C\C3=[N+](CCCCCC(O)=O)c4ccc5ccccc5c4C3(C)C)=C2Cl)C(C)(C)c2c1ccc1ccccc21

Application

Lipids Research

Packaging

5 mL Amber Glass Screw Cap Vial (810348P-5MG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service