Skip to Content
Merck

EHU131521

MISSION® esiRNA

targeting human COL1A2

Sign In to View Organizational & Contract Pricing.

Select a Size

EHU131521-20UG

€212.00

EHU131521-50UG

€382.00

€212.00


Check Cart for Availability


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CTCGCTCAGCACCTTCTCTCAGACCCAAGGACTATGAAGTTGATGCTACTCTGAAGTCTCTCAACAACCAGATTGAGACCCTTCTTACTCCTGAAGGCTCTAGAAAGAACCCAGCTCGCACATGCCGTGACTTGAGACTCAGCCACCCAGAGTGGAGCAGTGGTTACTACTGGATTGACCCTAACCAAGGATGCACTATGGATGCTATCAAAGTATACTGTGATTTCTCTACTGGCGAAACCTGTATCCGGGCCCAACCTGAAAACATCCCAGCCAAGAACTGGTATAGGAGCTCCAAGGACAAGAAACACGTCTGGCTAGGAGAAACTATCAATGCTGGCAGCCAGTTTGAATATAATGTAGAAGGAGTGACTTCCAAGGAAATGGCTACCCAACTTGCCTTCATGCGCCTGCTGGCCAACTATGCCTCTCAGAACATCACCTACCACTGCAAGAACAGCATTGCAT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
EHU153561EHU161021EHU025331
Ensembl | human accession no.

ENSG00000164692

Ensembl | human accession no.

ENSG00000108821

Ensembl | human accession no.

ENSG00000070729

Ensembl | human accession no.

ENSG00000137392

Gene Information

human ... COL1A2(1278), COL1A2(1278)

Gene Information

human ... COL1A1(1277), COL1A1(1277)

Gene Information

human ... CNGB1(1258), CNGB1(1258)

Gene Information

human ... CLPS(1208), CLPS(1208)

product line

MISSION®

product line

MISSION®

product line

MISSION®

product line

MISSION®

esiRNA cDNA target sequence

CTCGCTCAGCACCTTCTCTCAGACCCAAGGACTATGAAGTTGATGCTACTCTGAAGTCTCTCAACAACCAGATTGAGACCCTTCTTACTCCTGAAGGCTCTAGAAAGAACCCAGCTCGCACATGCCGTGACTTGAGACTCAGCCACCCAGAGTGGAGCAGTGGTTACTACTGGATTGACCCTAACCAAGGATGCACTATGGATGCTATCAAAGTATACTGTGATTTCTCTACTGGCGAAACCTGTATCCGGGCCCAACCTGAAAACATCCCAGCCAAGAACTGGTATAGGAGCTCCAAGGACAAGAAACACGTCTGGCTAGGAGAAACTATCAATGCTGGCAGCCAGTTTGAATATAATGTAGAAGGAGTGACTTCCAAGGAAATGGCTACCCAACTTGCCTTCATGCGCCTGCTGGCCAACTATGCCTCTCAGAACATCACCTACCACTGCAAGAACAGCATTGCAT

esiRNA cDNA target sequence

CTCCTGGCAAAGATGGACTCAACGGTCTCCCTGGCCCCATTGGGCCCCCTGGTCCTCGCGGTCGCACTGGTGATGCTGGTCCTGTTGGTCCCCCCGGCCCTCCTGGACCTCCTGGTCCCCCTGGTCCTCCCAGCGCTGGTTTCGACTTCAGCTTCCTGCCCCAGCCACCTCAAGAGAAGGCTCACGATGGTGGCCGCTACTACCGGGCTGATGATGCCAATGTGGTTCGTGACCGTGACCTCGAGGTGGACACCACCCTCAAGAGCCTGAGCCAGCAGATCGAGAACATCCGGAGCCCAGAGGGCAGCCGCAAGAACCCCGCCCGCACCTGCCGTGACCTCAAGATGTGCCACTCTGACTGGAAGAGTGGAGAGTACTGGATTGACCCCAACCAAGGCTGCAACCTGGATGCCATCAAAGTCTTCTGCAACATGGAGACTGGTGAGACCTGCGTGTACCCCACTCAGCCCAGTGTGGCCCAGAAGAACTGGTACATCAGCAAGAACCCCAAGGACAAGAGGCATGTCTGGTTCGGCGAGAGCATGACCGATGGATT

esiRNA cDNA target sequence

AGTGGAACCAGAGCCAGAGATGGAGGCGGAGGTGGAACCAGAACCGAATCCTGAGGAGGCCGAGACAGAGTCCGAGTCCATGCCCCCCGAAGAGTCATTCAAGGAGGAGGAAGTGGCTGTGGCAGACCCAAGCCCTCAGGAGACCAAGGAGGCTGCCCTTACTTCCACCATATCCCTCCGGGCCCAGGGCGCTGAGATTTCTGAAATGAATAGTCCCAGCCGCAGGGTACTGACCTGGCTCATGAAGGGCGTAGAGAAGGTGATCCCGCAGCCTGTTCACAGCATCACGGAGGACCCGGCTCAGATCCTGGGGCATGGCAGCACTGGGGACACAGGGTGCACAGATGAACCCAATGAGGCCCTTGAGGCCCAAGACACTAGGCCTGGGCTGCGGCTGCTTCTGTGGCTGGAGCAGAATCT

esiRNA cDNA target sequence

GATCCTGATCCTCCTGCTTGTCGCCCTCTCTGTGGCCTATGCAGCTCCTGGCCCCCGGGGGATCATTATCAACCTGGAGAACGGTGAGCTCTGCATGAATAGTGCCCAGTGTAAGAGCAATTGCTGCCAGCATTCAAGTGCGCTGGGCCTGGCCCGCTGCACATCCATGGCCAGCGAGAACAGCGAGTGCTCTGTCAAGACGCTCTATGGGATTTACTACAAGTGTCCCTGTGAGCGTGGCCTGACCTGTGAGGGAGACAAGACCATCGTGGGCTCCATCACCAACACCAACTTTGGCATCTGCCATGACGCTGGACGCTCCAAGCAGTGAGACTGCCCACCCACTCCCACACCTAGCCCAGAATGCTGTAGGCCACTAGGCGCAGGGGCATCTCTCCCCTGCTCCAGCGCATCTCCCGGGCTGGCCACCTCCTTGACCAGCATATCTGTTTTCTGATTGCGCTCTTCACAATTAAAGGCCTCCTGCAA

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

NCBI accession no.

NM_000089

NCBI accession no.

NM_000088

NCBI accession no.

NM_001297

NCBI accession no.

NM_001832

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xue-Feng Li et al.
Molecular therapy. Nucleic acids, 22, 1142-1153 (2020-12-10)
Dysregulated expression of microRNAs (miRNAs or miRs) has been implicated in the pathophysiology of type 2 diabetes mellitus (T2DM). However, their underlying role in the complication of detrusor fibrosis remains poorly understood. Therefore, this study aimed to examine the potential

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service