Merck
  • Berberine up-regulates hepatic low-density lipoprotein receptor through Ras-independent but AMP-activated protein kinase-dependent Raf-1 activation.
Berberine up-regulates hepatic low-density lipoprotein receptor through Ras-independent but AMP-activated protein kinase-dependent Raf-1 activation.
Biological & pharmaceutical bulletin (2014-09-10)
Zheng Li, Jian-Dong Jiang, Wei-Jia Kong
ABSTRACT
Our previous studies showed that berberine (BBR) increases liver low-density lipoprotein (LDL) receptor expression in an extracellular signal-regulated kinase (ERK)-dependent manner. This study was designed to explore the upstream cellular signaling molecules recruited by BBR to activate the ERK mitogen-activated protein kinase (MAPK) cascade. Chemical inhibitors such as GW5074, manumycin A, and compound C or specific small interfering RNAs (siRNAs) were used in the blocking experiments; Western blot was used to determine the phosphorylation of kinases; real-time reverse transcriptase polymerase chain reaction (RT-PCR) was used to determine the expression level of LDL receptor mRNA. Our results indicate that BBR increases p-Raf-1 (ser338) level time and dose dependently in HL-7702 cells, but has no influence on Ras activity; the stimulating activities of BBR on Raf-1 signaling and LDL receptor expression can be blocked by GW5074 completely, but not by manumycin A, a Ras inhibitor. BBR activates hepatic Raf-1 signaling and up-regulates LDL receptor expression in a rat model of hyperlipidemia with no impact on liver Ras activity. Importantly, our results show that the stimulating activities of BBR on hepatic Raf-1 signaling and LDL receptor expression are totally blocked by compound C, a selective inhibitor of AMP-activated protein kinase (AMPK), and also by silencing its expression with siRNA. Taken together, our results demonstrate for the first time that BBR up-regulates LDL receptor expression through Ras-independent, but AMPK-dependent Raf-1 activation in liver cells. Our study will help to elucidate the molecular pharmacology of BBR and provide new scientific evidence for its clinical application.
Product Description
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC), Molecular Biology
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Supelco
Staurosporine, VETRANAL®, analytical standard
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
GW5074, powder
Sigma-Aldrich
Deoxycholic acid, ≥99.0% (T)
Sigma-Aldrich
Deoxycholic acid, ≥98% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard