Merck
Isolation, culture, and immunostaining of neonatal rat ventricular myocytes.
STAR protocols (2021-11-26)
Ana Helena Macedo Pereira, Alisson Campos Cardoso, Kleber Gomes Franchini
RESUMEN
Isolation and culture of ventricular cardiomyocytes from neonatal rats (NRVMs) is a powerful model to study neonatal cardiac development, cell cycle regulation, and cardiac physiology and pathology in vitro. Here, we present our modified enzymatic digestion protocol followed by two-step discontinuous Percoll gradient centrifugation to isolate a high yield of viable ventricular cardiomyocytes from neonatal rats. Finally, here we describe an immunostaining protocol for cytosolic and nuclear staining of NRVMs. For complete details on the use and execution of this protocol, please refer to Pereira et al. (2020).
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Glicina, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Pancreatina from porcine pancreas, 4 × USP specifications
Sigma-Aldrich
Anti-α-actinina (sarcomérica) monoclonal antibody produced in mouse, clone EA-53, ascites fluid
Sigma-Aldrich
Seroalbúmina bovina, lyophilized powder, ≥98% (agarose gel electrophoresis)