- A graphene oxide-photosensitizer complex as an enzyme-activatable theranostic agent.
A graphene oxide-photosensitizer complex as an enzyme-activatable theranostic agent.
Chemical communications (Cambridge, England) (2013-01-04)
Youngnam Cho, Hyunjin Kim, Yongdoo Choi
PMID23283113
RESUMEN
The development of a target-activatable "smart" platform with high-affinity and specificity is of great importance for a wide range of biomedical applications. Here, we attempted to develop a new enzyme-activatable graphene oxide-photosensitizer complex as a biologically tunable theranostic agent, making it promising for near-infrared (NIR) fluorescence imaging and photo-induced cancer therapy.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Hialuronidasa from bovine testes, Type I-S, lyophilized powder, 400-1000 units/mg solid
Sigma-Aldrich
Hialuronidasa from bovine testes, Type IV-S, lyophilized powder (essentially salt-free), 750-3000 units/mg solid
Sigma-Aldrich
Hialuronidasa from bovine testes, Type IV-S, powder, suitable for mouse embryo cell culture, 750-3000 units/mg solid
Sigma-Aldrich
Hyaluronidase from sheep testes, Type V, lyophilized powder, ≥1,500 units/mg solid
Sigma-Aldrich
Hialuronidasa from bovine testes, Type VI-S, lyophilized powder, 3,000-15,000 units/mg solid