Merck
  • Identification of coregulators influenced by estrogen receptor subtype specific binding of the ER antagonists 4-hydroxytamoxifen and fulvestrant.
Identification of coregulators influenced by estrogen receptor subtype specific binding of the ER antagonists 4-hydroxytamoxifen and fulvestrant.
Chemico-biological interactions (2014-07-12)
Nynke M Evers, Si Wang, Johannes H J van den Berg, René Houtman, Diana Melchers, Laura H J de Haan, Antwan G H Ederveen, John P Groten, Ivonne M C M Rietjens
RESUMEN
The aim of the present study was to investigate modulation of the interaction of ERα and ERβ with coregulators in the ligand dependent responses induced by the ER antagonistic compounds 4OHT and fulvestrant. Comparison with the modulation index (MI) profiles for the ER agonist estradiol (E2) will elucidate whether differences in the (ant)agonist dependent interaction of ERα and ERβ with coregulators expressed in MI profiles contribute to the differences in (ant)agonist responses. To this end, the selected ER antagonistic compounds were first characterized for intrinsic relative potency and efficacy towards ERα and ERβ using ER selective U2OS reporter gene assays, and subsequently tested for ligand dependent modulation of the interaction of ERα and ERβ with coregulators using the MARCoNI assay. Results obtained indicate a preference of 4OHT to antagonize ERβ and find fulvestrant to be less ER specific. MARCoNI assay responses reveal that ERα and ERβ mediated interaction with coregulators expressed in MI profiles are similar for 4OHT and fulvestrant and generally opposite to the MI profile of the ER agonist E2. Hierarchical clustering based on the MI profiles appeared able to clearly discriminate the two compounds with ER antagonistic properties from the ER agonist E2. Taken together the data reveal that modulation of the interaction of ERs with coregulators discriminates ER agonists from antagonists but does not discriminate between the less specific ER antagonist fulvestrant and the preferential ERβ antagonistic compound 4OHT. It is concluded that differences in modulation of the interaction of ERα and ERβ with coregulators contribute to the differences in ligand dependent responses induced by ER agonists and ER antagonists but the importance of the subtle differences in modulation of the interaction of ERs with coregulators between the ER antagonistic compounds 4OHT and fulvestrant for the ultimate biological effect remains to be established.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Ácido clorhídrico, ACS reagent, 37%
Sigma-Aldrich
Hidróxido de sodio, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Hidróxido de sodio, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sulfato de magnesio, anhydrous, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Hidróxido de sodio solution, 50% in H2O
Sigma-Aldrich
Ácido clorhídrico, ACS reagent, 37%
Sigma-Aldrich
Cloruro de hidrógeno solution, 4.0 M in dioxane
Sigma-Aldrich
Hidróxido de sodio solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
DL-Ditiotreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Cloruro de sodio, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Hidróxido de sodio solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Ácido clorhídrico solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Cloruro de sodio, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Cloruro de sodio, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Hidróxido de sodio, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Hidróxido de sodio, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Supelco
DL-Ditiotreitol solution, 1 M in H2O
Sigma-Aldrich
Ácido clorhídrico, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Ácido clorhídrico, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Cloruro de hidrógeno solution, 2.0 M in diethyl ether
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Tricine, ≥99% (titration)
Sigma-Aldrich
Hidróxido de sodio, reagent grade, 97%, powder
Sigma-Aldrich
Sulfato de magnesio, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Ácido clorhídrico, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Hidróxido de sodio, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M