Merck
Fatty acid elongase-5 (Elovl5) regulates hepatic triglyceride catabolism in obese C57BL/6J mice.
Journal of lipid research (2014-05-13)
Sasmita Tripathy, Kelli A Lytle, Robert D Stevens, James R Bain, Christopher B Newgard, Andrew S Greenberg, Li-Shin Huang, Donald B Jump
RESUMEN
Nonalcoholic fatty liver disease is a major public health concern in the obese and type 2 diabetic populations. The high-fat lard diet induces obesity and fatty liver in C57BL/6J mice and suppresses expression of the PPAR-target gene, FA elongase 5 (Elovl5). Elovl5 plays a key role in MUFA and PUFA synthesis. Increasing hepatic Elovl5 activity in obese mice lowered hepatic TGs and endoplasmic reticulum stress markers (X-box binding protein 1 and cAMP-dependent transcription factor 6α) and increased TG catabolism and fatty acyl carnitines. Increased hepatic Elovl5 activity did not increase hepatic capacity for β-oxidation. Elovl5 effects on hepatic TG catabolism were linked to increased protein levels of adipocyte TG lipase (ATGL) and comparative gene identification 58 (CGI58). Elevated hepatic Elovl5 activity also induced the expression of some (pyruvate dehydrogenase kinase 4 and fibroblast growth factor 21), but not other cytochrome P450 4A10 (CYP4A10), PPAR-target genes. FA products of Elovl5 activity increased ATGL, but not CGI58, mRNA through PPARβ-dependent mechanisms in human HepG2 cells. Treatment of mouse AML12 hepatocytes with the PPARβ agonist (GW0742) decreased (14)C-18:2,n-6 in TGs but did not affect β-oxidation. These studies establish that Elovl5 activity regulates hepatic levels of FAs controlling PPARβ activity, ATGL expression, and TG catabolism, but not FA oxidation.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
HEPES, ≥99.5% (titration)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
(+)-Sodium L-ascorbate, crystalline, ≥98%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Sodium selenite, 99%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Sodium selenite, BioReagent, suitable for cell culture, ≥98%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
(+)-Sodium L-ascorbate, BioXtra, ≥99.0% (NT)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
(+)-Sodium L-ascorbate, powder, BioReagent, suitable for cell culture
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Sodium selenite, γ-irradiated, lyophilized powder, BioXtra, suitable for cell culture
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Supelco
Sodium ascorbate, Pharmaceutical Secondary Standard; Certified Reference Material
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Sodium selenite, anhydrous, ≥90.0% (RT)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sodium ascorbate, European Pharmacopoeia (EP) Reference Standard
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
En este momento no podemos mostrarle ni los precios ni la disponibilidad