Merck
  • Aspirin inhibits cell viability and mTOR downstream signaling in gastroenteropancreatic and bronchopulmonary neuroendocrine tumor cells.
Aspirin inhibits cell viability and mTOR downstream signaling in gastroenteropancreatic and bronchopulmonary neuroendocrine tumor cells.
World journal of gastroenterology (2014-08-12)
Matilde Spampatti, George Vlotides, Gerald Spöttl, Julian Maurer, Burkhard Göke, Christoph J Auernhammer
RESUMEN
To investigate the effect of aspirin on neuroendocrine tumor (NET) cell growth and signaling in vitro. Human pancreatic BON1, bronchopulmonary NCI-H727 and midgut GOT1 neuroendocrine tumor cells were treated with different concentrations of aspirin (from 0.001 to 5 mmol/L), and the resulting effects on metabolic activity/cell proliferation were measured using cell proliferation assays and SYBR-DNA-labeling after 72, 144 and 216 h of incubation. The effects of aspirin on the expression and phosphorylation of several critical proteins that are involved in the most common intracellular growth factor signaling pathways (especially Akt protein kinase B) and mammalian target of rapamycin (mTOR) were determined by Western blot analyses. Propidium iodide staining and flow cytometry were used to evaluate changes in cell cycle distribution and apoptosis. Statistical analysis was performed using a 2-tailed Student's t-test to evaluate the proliferation assays and cell cycle analyses. The results are expressed as the mean ± SD of 3 or 4 independently performed experiments. Statistical significance was set at P < 0.05. Treatment with aspirin suppressed the viability/proliferation of BON1, NCI-H727 and GOT1 cells in a time- and dose-dependent manner. Significant effects were observed at starting doses of 0.5-1 mmol/L and peaked at 5 mmol/L. For instance, after treatment with 1 mmol/L aspirin for 144 h, the viability of pancreatic BON1 cells decreased to 66% ± 13% (P < 0.05), the viability of bronchopulmonary NCI-H727 cells decreased to 53% ± 8% (P < 0.01) and the viability of midgut GOT1 cells decreased to 89% ± 6% (P < 0.01). These effects were associated with a decreased entry into the S phase, the induction of the cyclin-dependent kinase inhibitor p21 and reduced expression of cyclin-dependent kinase 4 and cyclin D3. Aspirin suppressed mTOR downstream signaling, evidenced by the reduced phosphorylation of the mTOR substrates 4E binding protein 1, serine/threonine kinase P70S6K and S6 ribosomal protein and inhibited glycogen synthase kinase 3 activity. We observed the (compensatory) activation of tuberous sclerosis 2, the serine/threonine specific protein kinase AKT and extracellular signal-regulated kinases. Aspirin demonstrates promising anticancer properties for NETs in vitro. Further preclinical and clinical studies are needed.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
L-Threonine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Supelco
Aspirin (Acetyl Salicylic Acid), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Aspirin, meets USP testing specifications
Sigma-Aldrich
L-Threonine, reagent grade, ≥98% (HPLC)
SAFC
L-Threonine
USP
Aspirin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Acetylsalicylic acid, analytical standard
Sigma-Aldrich
L-Threonine, BioXtra, ≥99.5% (NT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Supelco
L-Threonine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
DL-Serine, ≥98% (TLC)
Acetylsalicylic acid, European Pharmacopoeia (EP) Reference Standard
Acetylsalicylic acid for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
DL-Serine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98% (HPLC)
Serine, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Threonine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
L-Threonine, European Pharmacopoeia (EP) Reference Standard