Merck
Spontaneous transient hyperpolarizations in the rabbit small intestine.
The Journal of physiology (2014-09-14)
Yoshihiko Kito, Masaaki Kurahashi, Retsu Mitsui, Sean M Ward, Kenton M Sanders
RESUMEN
Four types of electrical activity were recorded and related to cell structure by intracellular recording and dye injection into impaled cells in muscles of rabbit small intestine. The specific cell types from which recordings were made were longitudinal smooth muscle cells (LSMCs), circular smooth muscle cells (CSMCs), interstitial cells of Cajal distributed in the myenteric region (ICC-MY) and fibroblast-like cells (FLCs). Slow waves (slow wavesSMC) were recorded from LSMCs and CSMCs. Slow waves (slow wavesICC) were of greatest amplitude (>50 mV) and highest maximum rate of rise (>10 V s(-1)) in ICC-MY. The dominant activity in FLCs was spontaneous transient hyperpolarizations (STHs), with maximum amplitudes above 30 mV. STHs were often superimposed upon small amplitude slow waves (slow wavesFLC). STHs displayed a cyclical pattern of discharge irrespective of background slow wave activity. STHs were inhibited by MRS2500 (3 μm), a P2Y1 antagonist, and abolished by apamin (0.3 μm), a blocker of small conductance Ca(2+)-activated K(+) channels. Small amplitude STHs (<15 mV) were detected in smooth muscle layers, whereas STHs were not resolved in cells identified as ICC-MY. Electrical field stimulation evoked purinergic inhibitory junction potentials (IJPs) in CSMCs. Purinergic IJPs were not recorded from ICC-MY. These results suggest that FLCs may regulate smooth muscle excitability in the rabbit small intestine via generation of rhythmic apamin-sensitive STHs. Stimulation of P2Y1 receptors modulates the amplitudes of STHs. Our results also suggest that purinergic inhibitory motor neurons regulate the motility of the rabbit small intestine by causing IJPs in FLCs that conduct to CSMCs.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Yoduro de propidio, ≥94.0% (HPLC)
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Nifedipine, ≥98% (HPLC), powder
Sigma-Aldrich
Yoduro de propidio, ≥94% (HPLC)
Sigma-Aldrich
Apamin, from bee venom, ≥95% (HPLC)
Sigma-Aldrich
Nω-Nitro-L-arginine, ≥98% (TLC)
Supelco
Nifedipine, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Nifedipine, United States Pharmacopeia (USP) Reference Standard
Atropine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Apamin, synthetic, ≥97% (HPLC)
Atropine for peak identification, European Pharmacopoeia (EP) Reference Standard
Nifedipine, European Pharmacopoeia (EP) Reference Standard