Merck
  • Effects of pathogenic mutations in membrane subunits of mitochondrial Complex I on redox activity and proton translocation studied by modeling in Escherichia coli.
Effects of pathogenic mutations in membrane subunits of mitochondrial Complex I on redox activity and proton translocation studied by modeling in Escherichia coli.
Mitochondrion (2015-03-10)
Jukka Pätsi, Marko Kervinen, Laura Kytövuori, Kari Majamaa, Ilmo E Hassinen
RESUMEN
Effects of Complex I mutations were studied by modeling in NuoH, NuoJ or NuoK subunits of Escherichia coli NDH-1 by simultaneous optical monitoring of deamino-NADH oxidation and proton translocation and fitting to the data a model equation of transmembrane proton transport. A homolog of the ND1-E24 LHON/MELAS mutation caused 95% inhibition of d-NADH oxidation and proton translocation. The NuoJ-Y59F replacement decreased proton translocation. The NuoK-E72Q mutation lowered the enzyme activity, but proton pumping could be rescued by the double mutation NuoK-E72Q/I39D. Moving the NuoK-E72/E36 pair one helix turn towards the periplasm did not affect redox activity but decreased proton pumping.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
MOPS, ≥99.5% (titration)
Sigma-Aldrich
DL-Ditiotreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
MES, low moisture content, ≥99% (titration)
Supelco
DL-Ditiotreitol solution, 1 M in H2O
Sigma-Aldrich
Fluoruro de fenilmetansulfonilo, ≥98.5% (GC)
Sigma-Aldrich
MOPS, BioPerformance Certified, suitable for cell culture, ≥99.5% (titration)
Sigma-Aldrich
Carbonyl cyanide 3-chlorophenylhydrazone, ≥97% (TLC), powder
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Fluoruro de fenilmetansulfonilo, ≥99.0% (T)
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Hidróxido de potasio, ≥99.95% trace metals basis
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
MES solution, BioUltra, Molecular Biology, 0.5 M in H2O
Sigma-Aldrich
Potassium thiocyanate, BioXtra, ≥99.0%
Sigma-Aldrich
Ethanesulfonic acid, 95%
Sigma-Aldrich
MOPS, BioXtra, ≥99.5% (titration)
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
MOPS, anhydrous, free-flowing, Redi-Dri, ≥99.5%