Merck
  • Regulation at multiple levels control the expression of folate transporters in liver cells in conditions of ethanol exposure and folate deficiency.
Regulation at multiple levels control the expression of folate transporters in liver cells in conditions of ethanol exposure and folate deficiency.
BioFactors (Oxford, England) (2015-07-15)
Shilpa Thakur, Jyotdeep Kaur
RESUMEN
Complex regulatory mechanisms control the expression of folate transporters within cells. Liver is the primary reserve of the folate stores within the body. As excessive alcohol consumption or inefficient dietary folate intake are known to create folate deficiency, so therefore the current study was designed to explore various regulatory mechanisms controlling the expression of folate transport in liver cells in conditions of ethanol exposure and folate deficiency. In order to see whether the effects mediated by the treatments are reversible or not, ethanol removal, and folate repletion was done after ethanol exposure and folate deficiency treatment respectively. Folate deficiency resulted an increase, whereas ethanol treatment decreased the folic acid uptake within the cells. The alterations in folic acid uptake were in agreement with the observed changes in the expression of folate transporters. Ethanol exposure resulted an increase in promoter methylation of reduced folate carrier; however, folate deficiency had no effect. The effects produced by ethanol exposure and folate deficiency were found to be reversible in nature as depicted in case of ethanol removal and folate repletion group. Rate of synthesis of folate transporters was found to be increased whereas half lives of mRNA of folate transporters was found to be decreased on folate deficiency treatment and reverse was the case on ethanol treatment. Overall, alteration in the expression of folate transporters under ethanol exposure and folate deficient conditions can be attributed to those regulatory mechanisms which work at the mRNA level.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Sacarosa, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Bicarbonato de sodio, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sacarosa, ≥99.5% (GC)
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Cloruro de sodio, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Sacarosa, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sacarosa, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Cloruro de sodio, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Cloruro de sodio, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sacarosa, ≥99.5% (GC)
Sigma-Aldrich
Sacarosa, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Hemin, from bovine, ≥90%
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Sacarosa, ACS reagent
Sigma-Aldrich
Hemin, BioXtra, from Porcine, ≥96.0% (HPLC)
Sigma-Aldrich
Thiamine pyrophosphate, ≥95%
Sigma-Aldrich
Sulfato de magnesio, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Cloruro de sodio, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Cloruro de sodio, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Cloruro de sodio, 99.999% trace metals basis
Sigma-Aldrich
Sacarosa, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Bicarbonato de sodio, BioXtra, 99.5-100.5%