Merck
  • In vitro solubility, dissolution and permeability studies combined with semi-mechanistic modeling to investigate the intestinal absorption of desvenlafaxine from an immediate- and extended release formulation.
In vitro solubility, dissolution and permeability studies combined with semi-mechanistic modeling to investigate the intestinal absorption of desvenlafaxine from an immediate- and extended release formulation.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences (2015-06-21)
F Franek, A Jarlfors, F Larsen, P Holm, B Steffansen
RESUMEN
Desvenlafaxine is a biopharmaceutics classification system (BCS) class 1 (high solubility, high permeability) and biopharmaceutical drug disposition classification system (BDDCS) class 3, (high solubility, poor metabolism; implying low permeability) compound. Thus the rate-limiting step for desvenlafaxine absorption (i.e. intestinal dissolution or permeation) is not fully clarified. The aim of this study was to investigate whether dissolution and/or intestinal permeability rate-limit desvenlafaxine absorption from an immediate-release formulation (IRF) and Pristiq(®), an extended release formulation (ERF). Semi-mechanistic models of desvenlafaxine were built (using SimCyp(®)) by combining in vitro data on dissolution and permeation (mechanistic part of model) with clinical data (obtained from literature) on distribution and clearance (non-mechanistic part of model). The model predictions of desvenlafaxine pharmacokinetics after IRF and ERF administration were compared with published clinical data from 14 trials. Desvenlafaxine in vivo dissolution from the IRF and ERF was predicted from in vitro solubility studies and biorelevant dissolution studies (using the USP3 dissolution apparatus), respectively. Desvenlafaxine apparent permeability (Papp) at varying apical pH was investigated using the Caco-2 cell line and extrapolated to effective intestinal permeability (Peff) in human duodenum, jejunum, ileum and colon. Desvenlafaxine pKa-values and octanol-water partition coefficients (Do:w) were determined experimentally. Due to predicted rapid dissolution after IRF administration, desvenlafaxine was predicted to be available for permeation in the duodenum. Desvenlafaxine Do:w and Papp increased approximately 13-fold when increasing apical pH from 5.5 to 7.4. Desvenlafaxine Peff thus increased with pH down the small intestine. Consequently, desvenlafaxine absorption from an IRF appears rate-limited by low Peff in the upper small intestine, which "delays" the predicted time to the maximal plasma concentration (tmax), consistent with clinical data. Conversely, desvenlafaxine absorption from the ERF appears rate-limited by dissolution due to the formulation, which tends to negate the influence of pH-dependent permeability on absorption. We suggest that desvenlafaxine Peff is mainly driven by transcellular diffusion of the unionized form. In the case of desvenlafaxine, poor metabolism does not imply low intestinal permeability, as indicated by the BDDCS, merely low duodenal/jejunal permeability.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Base Trizma®, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Bicarbonato de sodio, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Base Trizma®, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Tris(hidroximetil)-aminometano, ACS reagent, ≥99.8%
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), powder or crystals
Sigma-Aldrich
Base Trizma®, BioUltra, Molecular Biology, ≥99.8% (T)
Sigma-Aldrich
Base Trizma®, ≥99.0% (T)
Sigma-Aldrich
Base Trizma®, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Potassium, chunks (in mineral oil), 98% trace metals basis
Sigma-Aldrich
Bicarbonato de sodio, BioXtra, 99.5-100.5%
Sigma-Aldrich
Base Trizma®, ≥99.9% (titration), crystalline
Sigma-Aldrich
Base Trizma®, anhydrous, free-flowing, Redi-Dri, ≥99.9%
Sigma-Aldrich
Decylamine, 95%
Sigma-Aldrich
Potassium hydride, 30 wt % dispersion in mineral oil
Sigma-Aldrich
Base Trizma®, 99.7% (T), puriss. p.a.
Sigma-Aldrich
Tetraborato de sodio decahydrate, BioXtra, ≥99.5%
Sigma-Aldrich
Decylamine, ≥99.0% (GC)
Sigma-Aldrich
Potassium hydride, in paraffin
SAFC
Tromethamine
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
5-Norbornene-2-endo,3-exo-dicarboxylic acid, 97%