Merck
Flightless I interacts with NMMIIA to promote cell extension formation, which enables collagen remodeling.
Molecular biology of the cell (2015-04-17)
Pamma D Arora, Yongqiang Wang, Anne Bresnick, Paul A Janmey, Christopher A McCulloch
RESUMEN
We examined the role of the actin-capping protein flightless I (FliI) in collagen remodeling by mouse fibroblasts. FliI-overexpressing cells exhibited reduced spreading on collagen but formed elongated protrusions that stained for myosin10 and fascin and penetrated pores of collagen-coated membranes. Inhibition of Cdc42 blocked formation of cell protrusions. In FliI-knockdown cells, transfection with constitutively active Cdc42 did not enable protrusion formation. FliI-overexpressing cells displayed increased uptake and degradation of exogenous collagen and strongly compacted collagen fibrils, which was blocked by blebbistatin. Mass spectrometry analysis of FliI immunoprecipitates showed that FliI associated with nonmuscle myosin IIA (NMMIIA), which was confirmed by immunoprecipitation. GFP-FliI colocalized with NMMIIA at cell protrusions. Purified FliI containing gelsolin-like domains (GLDs) 1-6 capped actin filaments efficiently, whereas FliI GLD 2-6 did not. Binding assays showed strong interaction of purified FliI protein (GLD 1-6) with the rod domain of NMMIIA (kD = 0.146 μM), whereas FliI GLD 2-6 showed lower binding affinity (kD = 0.8584 μM). Cells expressing FliI GLD 2-6 exhibited fewer cell extensions, did not colocalize with NMMIIA, and showed reduced collagen uptake compared with cells expressing FliI GLD 1-6. We conclude that FliI interacts with NMMIIA to promote cell extension formation, which enables collagen remodeling in fibroblasts.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
2- Mercaptoetanol, Molecular Biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
2- Mercaptoetanol, ≥99.0%
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Cloruro de calcio solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
L-Glutatión reducido, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutatión reducido, ≥98.0%
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
2- Mercaptoetanol, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Cloruro de calcio dihydrate
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
L-Glutatión reducido, BioXtra, ≥98.0%
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Batimastat, ≥98% (HPLC)
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O