Merck
  • Differences in the Epigenetic Regulation of Cytochrome P450 Genes between Human Embryonic Stem Cell-Derived Hepatocytes and Primary Hepatocytes.
Differences in the Epigenetic Regulation of Cytochrome P450 Genes between Human Embryonic Stem Cell-Derived Hepatocytes and Primary Hepatocytes.
PloS one (2015-07-16)
Han-Jin Park, Young-Jun Choi, Ji Woo Kim, Hang-Suk Chun, Ilkyun Im, Seokjoo Yoon, Yong-Mahn Han, Chang-Woo Song, Hyemin Kim
RESUMEN
Human pluripotent stem cell-derived hepatocytes have the potential to provide in vitro model systems for drug discovery and hepatotoxicity testing. However, these cells are currently unsuitable for drug toxicity and efficacy testing because of their limited expression of genes encoding drug-metabolizing enzymes, especially cytochrome P450 (CYP) enzymes. Transcript levels of major CYP genes were much lower in human embryonic stem cell-derived hepatocytes (hESC-Hep) than in human primary hepatocytes (hPH). To verify the mechanism underlying this reduced expression of CYP genes, including CYP1A1, CYP1A2, CYP1B1, CYP2D6, and CYP2E1, we investigated their epigenetic regulation in terms of DNA methylation and histone modifications in hESC-Hep and hPH. CpG islands of CYP genes were hypermethylated in hESC-Hep, whereas they had an open chromatin structure, as represented by hypomethylation of CpG sites and permissive histone modifications, in hPH. Inhibition of DNA methyltransferases (DNMTs) during hepatic maturation induced demethylation of the CpG sites of CYP1A1 and CYP1A2, leading to the up-regulation of their transcription. Combinatorial inhibition of DNMTs and histone deacetylases (HDACs) increased the transcript levels of CYP1A1, CYP1A2, CYP1B1, and CYP2D6. Our findings suggest that limited expression of CYP genes in hESC-Hep is modulated by epigenetic regulatory factors such as DNMTs and HDACs.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Dimetilsulfóxido, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Dimetilsulfóxido, Molecular Biology
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Dimetilsulfóxido, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Triton X-100, laboratory grade
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Dimetilsulfóxido, ≥99.5% (GC), suitable for plant cell culture
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Dexametasona, powder, BioReagent, suitable for cell culture, ≥97%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
DAPI, for nucleic acid staining
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Formaldehído solution, Molecular Biology, 36.5-38% in H2O
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Formaldehído solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Dimetilsulfóxido, BioUltra, Molecular Biology, ≥99.5% (GC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Sodium butyrate, 98%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Dexametasona, ≥98% (HPLC), powder
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Formaldehído solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Dimetilsulfóxido, meets EP testing specifications, meets USP testing specifications
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Sodium butyrate, ≥98.5% (GC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Formaldehído solution, meets analytical specification of USP, ≥34.5 wt. %
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Dimetilsulfóxido, PCR Reagent
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Urea, suitable for electrophoresis
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Dexametasona, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
4-Aminosalicylic acid, 99%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Urea, meets USP testing specifications
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
En este momento no podemos mostrarle ni los precios ni la disponibilidad