About This Item
Skip To
biological source
mouse
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
DHIC3-5H10, monoclonal
species reactivity
human
packaging
antibody small pack of 25 μg
technique(s)
immunofluorescence: suitable
immunohistochemistry: suitable (paraffin)
isotype
IgMκ
target post-translational modification
unmodified
Related Categories
1 of 4
This Item | MABS2045 | MABS1998 | MABS1999 |
|---|---|---|---|
| species reactivity human | species reactivity human | species reactivity human | species reactivity human |
| clone DHIC3-5H10, monoclonal | clone DHIC2-2B4, monoclonal | clone HIC3-2D12, monoclonal | clone HIC1-2B4, monoclonal |
| antibody form purified immunoglobulin | antibody form purified antibody | antibody form purified immunoglobulin | antibody form purified immunoglobulin |
| biological source mouse | biological source mouse | biological source mouse | biological source mouse |
| technique(s) immunofluorescence: suitable, immunohistochemistry: suitable (paraffin) | technique(s) flow cytometry: suitable, immunohistochemistry: suitable (paraffin) | technique(s) flow cytometry: suitable, immunohistochemistry: suitable (paraffin) | technique(s) flow cytometry: suitable, immunofluorescence: suitable, immunohistochemistry: suitable (paraffin) |
| isotype IgMκ | isotype IgG1λ | isotype IgMκ | isotype IgG2bκ |
General description
Immunogen
Application
Signaling
Biochem/physiol Actions
Physical form
Preparation Note
Analysis Note
Immunohistochemistry (Paraffin) Analysis: A 1:50 dilution of this antibody detected HPd2 in human pancreatic tissue sections.
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service


