Skip to Content
Merck

Skip To

BPS79259

BPS Medium 1

Synonym(s):

BPS Medium

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
41116155
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


manufacturer/tradename

BPS Bioscience

storage temp.

2-8°C

General description

This liquid BPS Medium is optimized for use with many BPS Reporter Kits including:
  • 60612: AP1 Reporter Kit (JNK Pathway)
  • 60514: ARE Reporter Kit (Nrf2 Antioxidant Pathway)
  • 60611: CRE/CREB Reporter Kit (cAMP/PKA Signaling Pathway)
  • 60614: NF-κB Reporter Kit (NF-κB Signaling Pathway)
  • 60509: Notch1/CSL Reporter Kit (Notch Signaling Pathway)
  • 60654: SBE Reporter Kit (TGFβ/SMAD signaling pathway)
  • 60511: SRE Reporter Kit (MAPK/ERK Signaling Pathway)
  • 60500: TCF/LEF Reporter Kit (Wnt Signaling Pathway)

Application

Datasheets and Protocols may be found here.

Preparation Note

Sterile filtered (0.2 µm) and Mycoplasma-free

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
BPS79260BPS7836643034
description

-

description

-

description

-

description

analytical standard

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

-65 to -96°C

storage temp.

-

manufacturer/tradename

BPS Bioscience

manufacturer/tradename

BPS Bioscience

manufacturer/tradename

BPS Bioscience

manufacturer/tradename

-


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Skin Sens. 1

Storage Class

10 - Combustible liquids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

10043-52-4

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library