Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
41116155
manufacturer/tradename
BPS Bioscience
storage temp.
2-8°C
General description
This liquid BPS Medium is optimized for use with select BPS reporter kits.
Application
Datasheet and Protocols may be found here.
Preparation Note
Sterile filtered (0.2 µm) and Mycoplasma-free
Signal Word
Warning
Hazard Codes
Precautionary Statements
Hazard Classifications
Skin Sens. 1
Storage Class
10 - Combustible liquids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
10043-52-4
CAS No.
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
