Skip to Content
Merck

ZRB3117

Anti-β2-Microglobulin Antibody, clone 1E4 ZooMAb® Rabbit Monoclonal

newenhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Beta-2-microglobulin, Beta-2-microglobulin form pI 5.3, CDABP0092, HDCMA22P

Sign In to View Organizational & Contract Pricing

Select a Size

Pricing and availability is not currently available.

About This Item

UNSPSC Code:
12352203

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1E4, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 13.72 kDa
observed mol wt ~13 kDa

purified by

using Protein A

species reactivity

human

species reactivity (predicted by homology)

monkey

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... B2M(567)

General description

Beta-2-microglobulin (UniProt: P61769; also known as Beta-2-microglobulin form pI 5.3) is encoded by the B2M (also known as CDABP0092, HDCMA22P) gene (Gene ID: 567) in human. β2-Microglobulin is a small protein characterized by its homology to immunoglobulin constant domains, particularly the CH3 domain of IgG molecules. It consists of a single polypeptide chain stabilized by a disulfide bond, contributing to its compact and stable structure. β2-Microglobulin is a critical component of the major histocompatibility complex (MHC) class I molecules, where it non-covalently associates with the heavy chain to facilitate antigen presentation to cytotoxic T cells. β2-Microglobulin is ubiquitously expressed on the surface of most nucleated cells and is also found in extracellular fluids due to its continual release during cellular turnover. β2-Microglobulin is localized to the plasma membrane as part of MHC-I complexes and can be internalized during endocytosis. It interacts with proteins such as FcRn (neonatal Fc receptor), influencing immune responses and protein recycling. It is critical for the immune surveillance and modulation in vertebrate animals. Dysregulation of β2-Microglobulin is implicated in diseases such as dialysis-related amyloidosis, bone and joint disorders and certain cancers, where its aggregation and altered expression contribute to pathological processes. It can cross the blood brain barrier, affecting certain CNS diseases. This ZooMAb® recombinant monoclonal antibody, generated by our proprietary technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Liu Z. Y., et al., (2024) Cell Mol Neurobiol. 44(1), 46; Wang, C., et al., (2022), Front. Oncol. 12, 944722; Batista Muñoz A., et al., (2019), PLoS ONE 14(5), e0216498).
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Immunogen

KLH-conjugated linear peptide corresponding to 16 amino acids from the N-terminal half of human β2-Microglobulin.

Application

Quality Control Testing

Evaluated by Western Blotting in U-937 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected β2-Microglobulin in U-937 cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected β2-Microglobulin in human spleen tissue lysate.

Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected β2-Microglobulin in human liver tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound with β2-Microglobulin peptide with at least one thousand-fold (1,000X) higher affinity than with non-specific control peptide.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Biochem/physiol Actions

Clone 1E4 is a ZooMAb® rabbit recombinant monoclonal antibody that detects β2-Microglobulin. It targets an epitope within 16 amino acids from the N-terminal half.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Preparation Note

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Other Notes

We are committed to bringing you greener alternative products, which adhere to one or more of the 12 Principles of Green Chemistry. This antibody is preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed, and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service