Merck
Lysosomal glycerophosphocholine phosphodiesterase in Tetrahymena.
Biochemistry and molecular biology international (1999-04-17)
J Florin-Christensen, M Florin-Christensen
RÉSUMÉ
The purification and characterization of a novel phosphodiesterase (PDE) is presented. The activity was detected in the extracellular medium of Tetrahymena thermophila cultures, by the release of p-nitrophenol from p-nitrophenylphosphocholine (PNPPC) with an acidic pH optimum. In cell homogenates, it is sedimentable, shows a latency similar to that of acid phosphatase and is co-secreted with this enzyme, indicating that it is a lysosomal hydrolase. PNPPC-PDE was purified to homogeneity from the extracellular medium, yielding a single band of 58 kD on SDS polyacrylamide gel electrophoresis. It catalyzed the release of glycerol from glycerophosphocholine (GPC) and GPC competitively inhibits degradation of PNPPC. We present further evidence indicating that the natural substrate for PNPPC-PDE is GPC. Thus, Tetrahymena becomes the first eukaryote in which a lysosomal GPC-PDE is observed. This finding provides a new pathway for the complete breakdown of phosphatidylcholine in a lysosomal medium.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
sn-Glycerol-3-phosphocholine Phosphodiesterase from mold, lyophilized powder, ≥5 units/mg protein