Merck
p-Hydroxyphenylacetic Acid Metabolism in Pseudomonas putida F6.
Journal of bacteriology (2001-02-24)
K E O'Connor, B Witholt, W Duetz
RÉSUMÉ
Pseudomonas putida F6 was found to metabolize p-hydroxyphenylacetic acid through 3,4-dihydroxyphenylacetic acid, 3,4-dihydroxymandelic acid, and 3,4-dihydroxybenzaldehyde. Cell extracts of P. putida F6 catalyze the NAD(P)H-independent hydroxylation of p-hydroxyphenylacetic acid to 3,4-dihydroxyphenylacetic acid which is further oxidized to 3,4-dihydroxymandelic acid. Oxidation and decarboxylation of the latter yields 3,4-dihydroxybenzaldehyde. A red-brown color accompanies all of the above enzyme activities and is probably due to the polymerization of quinone-like compounds. 3,4-Dihydroxybenzaldehyde is further metabolized through extradiol ring cleavage.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
DL-3,4-Dihydroxymandelic acid, 95%