Merck
Rifaximin for preventing acute graft-versus-host disease: impact on plasma markers of inflammation and T-cell activation.
Journal of pediatric hematology/oncology (2013-01-01)
Muna Qayed, Amelia Langston, Kuang-Yueh Chiang, Keith August, Joseph A Hilinski, Conrad R Cole, Andre Rogatko, Roberd M Bostick, John T Horan
RÉSUMÉ
In murine allogeneic hematopoietic cell transplantation models, inhibiting bacterial translocation stemming from conditioning-induced damage to the gut mucosa abrogates inflammatory stimulation of donor T cells, preventing acute graft-versus-host disease (AGVHD). We conducted a phase I trial to begin testing the hypothesis that rifaximin, a broadly acting oral antibiotic, would reduce systemic inflammation and T-cell activation. We administered rifaximin to 20 adolescents and younger adults (day -10 through day +30) receiving intensive conditioning. We measured the plasma level of interleukin-6, as a marker of conditioning-induced inflammation, and the levels of soluble tumor necrosis factor receptor-1 and soluble interleukin-2 receptor, as surrogate markers of AGVHD. We formed a historical control group (n=24), from a previous study of biomarkers in AGVHD. The increase in the treatment group's mean interleukin-6 level from baseline to day 0 was 73% less than that in the control group (P=0.006). The increase from baseline to day 15 in the treatment group's mean soluble tumor necrosis factor-1 and soluble interleukin-2 receptor levels was similar to the control group. Incidences of grade 2 to 4 AGVHD also did not differ. This suggests that rifaximin may abrogate bacterial translocation and resultant inflammation, but in alternative donor transplants this does not prevent downstream activation of donor T cells.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
Rifaximin
Supelco
Rifaximin, VETRANAL®, analytical standard