Merck
Prognostic utility of secretory phospholipase A(2) in patients with stable coronary artery disease.
Clinical chemistry (2011-07-26)
Michelle L O'Donoghue, Ziad Mallat, David A Morrow, Joelle Benessiano, Sarah Sloan, Torbjørn Omland, Scott D Solomon, Eugene Braunwald, Alain Tedgui, Marc S Sabatine
RÉSUMÉ
Secretory phospholipase A(2) (sPLA(2)) may contribute to atherogenesis. To date, few prospective studies have examined the utility of sPLA(2) for risk stratification in coronary artery disease (CAD). We measured plasma sPLA(2) activity at baseline in 3708 subjects in the PEACE randomized trial of trandolapril vs placebo in stable CAD. Median follow-up was 4.8 years. We used Cox regression to adjust for demographics, clinical risk factors, apolipoprotein B, apolipoprotein A1, and medications. After multivariable adjustment, sPLA(2) was associated with an increased risk of cardiovascular death, myocardial infarction, or stroke (adjusted hazard ratio Q4:Q1 1.55, 95% CI 1.13-2.14) and cardiovascular death or heart failure (1.91, 1.20-3.03). In further multivariable assessment, increased activity levels of sPLA(2) were associated with the risk of cardiovascular death, myocardial infarction, or stroke (adjusted hazard ratio 1.47, 95% CI 1.06-2.04), independent of lipoprotein-associated phospholipase A(2) mass and C-reactive protein, and modestly improved the area under the curve (AUC) beyond established clinical risk factors (AUC 0.668-0.675, P = 0.01). sPLA(2), N-terminal pro-B-type natriuretic peptide, and high-sensitivity cardiac troponin T all were independently associated with cardiovascular death or heart failure, and each improved risk discrimination (P = 0.02, P < 0.001, P < 0.001, respectively). sPLA(2) activity provides independent prognostic information beyond established risk markers in patients with stable CAD. These data are encouraging for studies designed to evaluate the role of sPLA(2) as a therapeutic target.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
Trandolapril, ≥98% (HPLC), white, powder