Merck
Requirement for the transcription factor LSIRF/IRF4 for mature B and T lymphocyte function.
Science (New York, N.Y.) (1997-01-24)
H W Mittrücker, T Matsuyama, A Grossman, T M Kündig, J Potter, A Shahinian, A Wakeham, B Patterson, P S Ohashi, T W Mak
RÉSUMÉ
Lymphocyte-specific interferon regulatory factor (LSIRF) (now called IRF4) is a transcription factor expressed only in lymphocytes. Mice deficient in IRF4 showed normal distribution of B and T lymphocyes at 4 to 5 weeks of age but developed progressive generalized lymphadenopathy. IRF4-deficient mice exhibited a profound reduction in serum immunoglobulin concentrations and did not mount detectable antibody responses. T lymphocyte function was also impaired in vivo; these mice could not generate cytotoxic or antitumor responses. Thus, IRF4 is essential for the function and homeostasis of both mature B and mature T lymphocytes.