Merck
NLRP3 is Required for Complement-Mediated Caspase-1 and IL-1beta Activation in ICH.
Journal of molecular neuroscience : MN (2016-12-10)
Sheng-Tao Yao, Fang Cao, Jia-Lin Chen, Wei Chen, Rui-Ming Fan, Gang Li, You-Chao Zeng, Song Jiao, Xiang-Ping Xia, Chong Han, Qi-Shan Ran
RÉSUMÉ
Complement-mediated inflammation plays a vital role in intracerebral hemorrhage (ICH), implicating pro-inflammatory factor interleukin-1beta (IL-1β) secretion. Brain samples and contralateral hemiencephalon were all collected and detected by Western blot. NLRP3 expression was located by dual immunofluorescence staining at 1, 3, and 5 days post-ICH. Brain water content was examined post-ICH. The neural deficit scores were evaluated by observers blindly. ILs were detected by ELISA. SiRNAs targeting NLRP3 (siNLRP3), siASC, and siControl were injected to inhibit NLRP3 function. To test the complement activation via Nod-like receptor (NLR) family pyrin domain-containing 3 (NLRP3), normal rabbit complement (NRC) was injected with lipopolysaccharide (LPS) to facilitate the complement function. As a result, complement 3a (C3a) and complement 5a (C5a) were upregulated during the ICH-induced neuroinflammation, and ablation of C3 attenuates ICH-induced IL-1β release. Though the LPS rescues the neuroinflammation in the ICH model, C3 deficiency attenuates the LPS-induced inflammatory effect. The NLRP3 inflammasome was activated after ICH and was located in the microglial cell of the mouse brain, which exhibits a time-dependent manner. However, the number of NLRP3/Iba-1 dual-labeled cells in the C3
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
MISSION® esiRNA, targeting human NLRP3