Merck
Palladium-catalyzed carbonylative annulation of o-alkynylphenols: syntheses of 2-substituted-3-aroyl-benzo[b]furans.
The Journal of organic chemistry (2002-04-02)
Youhong Hu, Yan Zhang, Zhen Yang, Reza Fathi
RÉSUMÉ
We report here a general synthetic methodology for palladium-catalyzed carbonylative annulation of o-alkynylphenol to construct 2-substituted-3-aroyl-benzo[b]furan. On the basis of the results, this methodology could be applied to a wider selection of iodide substrates to generate desired products. In accordance with mechanistic studies, this process involves coordination of cationic and less hindered acyl palladium complexes with o-alkynylphenols to create a desired cascade triad (coordination, nucleophilic addition, and reductive elimination). Consistent with this mechanism, addition of 1 equiv of AgBF(4) to palladium catalyst Pd(Ph(3)P)(4) generates an ideal candidate for this unique transformation.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
Silver tetrafluoroborate, 98%