Merck
  • Effect of structural analogs of butaclamol (a new antipsychotic drug) on striatal homovanillic acid and adenyl cyclase of olfactory tubercle in rats.
Effect of structural analogs of butaclamol (a new antipsychotic drug) on striatal homovanillic acid and adenyl cyclase of olfactory tubercle in rats.
Canadian journal of physiology and pharmacology (1976-08-01)
T A Pugsley, J Merker, W Lippman
PMID10057
RÉSUMÉ
The 3-isopropyl (I), 3-cyclohexyl (II) and 3-phenyl (III) analogs of the new antipsychotic drug butaclamol, which contains a 3-tertiary butyl group, and their respective (+)-enantiomers, but not (-)-enantiomers, caused a dose related elevation of rat striatal homovanillic acid concentration, indicative of an increased dopamine (DA) turnover; droperidol also exhibited this activity. The order of activity of the (+)-enantiomers was (butaclamol) approximately II greater than I greater than III. A decrease in striatal DA was observed with (+)-I and (+)-III at the highest dose used, but not at one-half the dose. Each analog antagonized the DA-induced increase in adenyl cyclase (EC 4.6.1.1) activity of olfactory tubercle homogenates, the order of activity of the racemates (except for II) AND (+)-ENANTIOMERS BEING (BUTACLAMOL) APPROXIMATELY I greater than III greater than II. The (+)-enantiomers of butaclamol and analogs were two to four times more potent than their respective racemates, with (+)-butaclamol and (+)-I displaying activity generally equivalent to fluphenazine. The respective (-)-enantiomers were ineffective indicating a stereochemical specificity for DA-receptor blockade. Such analogs presented should be of value in elucidating dopaminergic mechansims.
MATÉRIAUX
Numéro du produit
Marque
Description du produit
Sigma-Aldrich
(−)-Butaclamol hydrochloride, solid