Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Pricing and availability is not currently available. Contact Technical Service
Inchi Code
1S/C5H5ClN2O/c1-8-5(6)4(3-9)2-7-8/h2-3H,1H3
Inchi Key
BRTDPKPJDXNQEX-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
97%
Storage temp.
Inert atmosphere, 2-8C
Country of Origin
CN
Physical Form
Pale-yellow to Yellow-brown Solid
Still not finding the right product?
Explore all of our products under 5-Chloro-1-methyl-1H-pyrazole-4-carbaldehyde
hstatements
Signal Word
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
