Skip to Content
Merck

MABC1092

Anti-ASPP2 (TP53BP2) Antibody, clone DX54.10

clone DX54.10, from mouse

Synonym(s):

Apoptosis-stimulating of p53 protein 2, Bcl2-binding protein, Bbp, Renal carcinoma antigen NY-REN-51, Tumor suppressor p53-binding protein 2, 53BP2, p53-binding protein 2, p53BP2

Sign In to View Organizational & Contract Pricing

Select a Size

Pricing and availability is not currently available.

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

DX54.10, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μL

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... TP53BP2(7159)

General description

Apoptosis-stimulating of p53 protein 2 (UniProt: Q13625; also known as Bcl2-binding protein, Bbp, Renal carcinoma antigen NY-REN-51, Tumor suppressor p53-binding protein 2, 53BP2, p53-binding protein 2, p53BP2) is encoded by the TP53BP2 (also known as ASPP2, BBP) gene (Gene ID: 7159) in human. 53BP2 plays a central role in regulation of apoptosis and cell growth. It is widely expressed and its expression is observed in spleen, thymus, prostate, testis, ovary, small intestine, colon and peripheral blood leukocyte 53BP2 is induced following DNA damage by UV radiation. It is down-regulated by wild-type p53/TP53, but not by mutant p53/TP53. It regulates p53/TP53 action by enhancing the DNA binding and transactivation function of TP53 on the promoters of pro-apoptotic genes in vivo. 53BP2 is shown to inhibit the ability of APPBP1 to conjugate NEDD8 to CUL1, and thereby decreases APPBP1 ability to induce apoptosis. 53BP2 contains four ankyrin repeats and one SH3 domain are required for a specific interactions with TP53. The 53BP2 binding site on the p53 core domain consists of evolutionarily conserved regions that are frequently mutated in human cancers. Lower level of 53BP2 expression is observed in breast carcinomas expressing a wild-type TP53 protein.
~170 kDa observed; 125.62 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Immunogen

Epitope: C-terminus
GST-tagged recombinant fragment corresponding to 438 amino acids from the C-terminal half of human Apoptosis-stimulating of p53 protein 2 (TP53BP2).

Application

Anti-ASPP2 (TP53BP2), clone DX54.10, Cat. No. MABC1092, is a mouse monoclonal antibody that detects Apoptosis-stimulating of p53 protein 2 (ASPP2) and has been tested for use in Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, and Western Blotting.
Immunoprecipitation Analysis: A representative lot detected ASPP2 (TP53BP2) in Immunoprecipitation applications (Samuels-Lev, Y., et. al. (2001). Mol Cell. 8(4):781-94).

Immunohistochemistry Analysis: A representative lot detected ASPP2 (TP53BP2) in Immunohistochemistry applications (Wang, Y., et. al. (2014). Nat Cell Biol. 16(11):1092-104).

Immunocytochemistry Analysis: A representative lot detected ASPP2 (TP53BP2) in Immunocytohemistry applications (Wang, Y., et. al. (2013). Cell Death Differ. 20(4):525-34; Wang, Y., et. al. (2014). Nat Cell Biol. 16(11):1092-104).

Western Blotting Analysis: A representative lot detected ASPP2 (TP53BP2) in Western Blotting applications (Wang, Y., et. al. (2013). Cell Death Differ. 20(4):525-34; Samuels-Lev, Y., et. al. (2001). Mol Cell. 8(4):781-94).
Research Category
Apoptosis & Cancer

Biochem/physiol Actions

Clone DX54.10 detects Apotosis -stimulating of p53 protein 2 (TP53BP2) in human cells. It targets an epitope with in 438 amino acids from the C-terminal half.

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in U2OS-iASPP and U2OS RISC-free cell lysates.

Western Blotting Analysis: A 1:125 dilution of this antibody detected ASPP2 in lysates of U2OS cells transfected with siRNA to knock down iASPP and in RISC-free U2OS cells.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service