Skip to Content
Merck

Z744415

2mag MIXControl 40 control unit

new

for connecting up to 2 MIXdrive stirring devices, with RS232 interface, AC/DC input 100 - 240 V, UK plug

Synonym(s):

2Mag 90410, control device, control module, lab stirrer control device, lab stirrer control module

Sign In to View Organizational & Contract Pricing

Select a Size


About This Item

UNSPSC Code:
41103806
NACRES:
NB.43
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

material

stainless steel

feature

40 W stirring power
Protection class IP20

packaging

pack of 1 ea

manufacturer/tradename

2mag 90410

parameter

0-40 °C temp. range (at 80% humidity)
100 – 2,000 rpm speed

stirrer W × D × H

200 mm × 155 mm × 38 mm

compatibility

for connecting up to 2 MIXdrive stirring devices

Looking for similar products? Visit Product Comparison Guide

General description

Powerful 2mag control unit for the connection of up to two MIXdrive stirring drives, powerful 40 watts stirring power, wide speed range from 100 up to 2,000 rpm, jerk-free stirring also at low speeds, power setting (10-steps) for high power with regard to large and viscous stirring amounts and reduced power for the warming-free continuous operation of MIXdrive stirring drives. Available with or without RS232 interface.

Application

  • Chemical reactions
  • Cell culture
  • Microbiology.

Features and Benefits

  • 100-2,000 rpm
  • 10 power steps
  • 2 stirrer sockets
  • Clear digital display
  • SoftStart for reliable catching/centering and safe accelerating of the
  • stirring bar
  • QuickSet for quick setting of start- and maximum
  • speed
  • Automatic memory function
  • Easy to clean stainless steel


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service