Sign In to View Organizational & Contract Pricing
Select a Size
About This Item
Empirical Formula (Hill Notation):
C37H44Cl2N2O2
Molecular Weight:
619.66
UNSPSC Code:
12352212
NACRES:
NA.21
biological source
synthetic
Assay
>99% (TLC)
form
powder
packaging
pkg of 1 × 5 mg (810353P-5MG)
manufacturer/tradename
Avanti Research™ - A Croda Brand
lipid type
fluorescent lipids
shipped in
dry ice
storage temp.
−20°C
SMILES string
[Cl-].CN1\C(=C\C=C2/CCCC(\C=C\C3=[N+](CCCCCC(O)=O)c4ccccc4C3(C)C)=C2Cl)C(C)(C)c2ccccc12
Related Categories
Application
Lipids Research
Legal Information
Avanti Research is a trademark of Avanti Polar Lipids, LLC
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service