About This Item
Skip To
biological source
rabbit
Quality Level
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
purified by
affinity chromatography
species reactivity
human, rat
species reactivity (predicted by homology)
porcine (based on 100% sequence homology), equine (based on 100% sequence homology)
technique(s)
ChIP: suitable
immunocytochemistry: suitable
western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... CREB1(1385)
1 of 4
This Item | MAB5432 | SAB1306301 | SAB4500443 |
|---|---|---|---|
| biological source rabbit | biological source mouse | biological source rabbit | biological source rabbit |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| antibody form affinity isolated antibody | antibody form purified immunoglobulin | antibody form affinity isolated antibody | antibody form affinity isolated antibody |
| clone polyclonal | clone monoclonal | clone polyclonal | clone polyclonal |
| technique(s) ChIP: suitable, western blot: suitable, immunocytochemistry: suitable | technique(s) ELISA: suitable, western blot: suitable | technique(s) immunohistochemistry: 1:50-1:100, western blot: 1:1000 | technique(s) ELISA: 1:10000, immunohistochemistry: 1:50-1:100, western blot: 1:500-1:1000 |
| UniProt accession no. | UniProt accession no. | UniProt accession no. | UniProt accession no. |
General description
Immunogen
Application
Chromatin Immunoprecipitation (ChIP) Analysis: A representative lot detected CREB-1 in HEK293T cells.
Epigenetics & Nuclear Function
Transcription Factors
Biochem/physiol Actions
Physical form
Preparation Note
Analysis Note
A431 cell lysate
Western Blotting Analysis: 1 µg/mL of this antibody detected CREB-1 in 10 µg of A431 cell lysate.
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service


