Skip to Content
Merck

87860

Tetramethylhydrazine

≥97.0%

Sign In to View Organizational & Contract Pricing

Select a Size

Pricing and availability is not currently available.

About This Item

Linear Formula:
(CH3)2NN(CH3)2
CAS Number:
Molecular Weight:
88.15
Beilstein:
1731483
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Quality Level

Assay

≥97.0%

form

liquid

impurities

≤2% water

refractive index

n20/D 1.404

density

0.777 g/mL at 20 °C (lit.)

SMILES string

CN(C)N(C)C

InChI

1S/C4H12N2/c1-5(2)6(3)4/h1-4H3

InChI key

DHBZRQXIRAEMRO-UHFFFAOYSA-N

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

80.6 °F - closed cup

Flash Point(C)

27 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An electron spin resonance study of the novel radical cation produced during the horseradish peroxidase-catalyzed oxidation of tetramethylhydrazine.
B Kalyanaraman et al.
Biochemical and biophysical research communications, 105(1), 217-224 (1982-03-15)
P A Craven et al.
Biochemical pharmacology, 34(17), 3101-3106 (1985-09-01)
A fatty acid stimulated, NADPH-independent pathway for the N-demethylation of 1,1-dimethylhydrazine (1,1-DMH) with the generation of HCHO was demonstrated in 10,000 g soluble fractions of colonic mucosal homogenates. Tetramethylhydrazine and, to a lesser extent, aminopyrine, but not 1,2-DMH or methylhydrazine
B Kalyanaraman et al.
Carcinogenesis, 4(10), 1341-1343 (1983-10-01)
Prostaglandin endoperoxide synthase catalyzed the oxidation of tetramethylhydrazine to the tetramethylhydrazine radical cation, as detected by electron spin resonance spectroscopy. Oxidation of tetramethylhydrazine by prostaglandin endoperoxide synthase also resulted in the N-demethylation of tetramethylhydrazine leading to the formation of formaldehyde.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service