Skip to Content
Merck

CLS353037

Corning® Falcon® Cell Culture Dish

plate diam. × capacity 60 x 15 mm × 6 mL, Tissue Culture (TC)-treated surface, sterile

Synonym(s):

Cell Culture Dish

Sign In to View Organizational & Contract Pricing.

Select a Size

500 EA

₪3,505.00

₪3,505.00


Please contact Customer Service for Availability


About This Item

NACRES:
NB.14

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Corning® Falcon® Cell Culture Dish, plate diam. × capacity 60 x 15 mm × 6 mL, Tissue Culture (TC)-treated surface, sterile

sterility

sterile

packaging

case of 500 ea (25 packs of 20)

manufacturer/tradename

Corning 353037

plate diam. × capacity

60 x 15 mm × 6 mL

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

Application

Cell Culture

General description

Falcon® 60mm X 15mm Style Center Well Organ Culture Dish, Tissue Culture-Treated, Sterile, Bulk Packaged, 20/Pack, 500/Case

Legal Information

Corning is a registered trademark of Corning, Inc.
Falcon is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service